Search
Search

Slc6a1| Gene Symbol: Slc6a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178703.4 | NP_848818.1 | 14 - 15 | 1907 | 68 | |
| GenBank mRNA | AK042956.1 | - | 14 - 15 | 1769 | 68 | |
| AK052971.1 | - | 14 - 15 | 1732 | 68 | ||
| AK053883.1 | - | 14 - 15 | 1729 | 68 | ||
| BC027111.1 | - | 1 - 2 | 102 | 68 | ||
| BC059080.1 | - | 14 - 15 | 1758 | 6827121 | 68 | |
| M92378.1 | - | 13 - 14 | 1666 | 68 | ||
Set Membership: |
|