Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01192446_m1
          See other TMCO2 GE Assays ›
          Gene Symbol:
          Tmco2
          Gene Name:
          transmembrane and coiled-coil domains 2
          Gene Aliases:
          1700028F16Rik
          Chromosome Location:
          Chr.4: 121105651 - 121109226 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01192446_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmco2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081312.1 NP_001074781.1 1 - 2 311 142
          GenBank mRNA AK006456.1 - 1 - 2 312 142

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003453.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P482814 NM_001081312.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P482814 AK006456.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P482814 NM_001081312.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P482814 AK006456.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 143989 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 13552 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152127 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P482814 NM_001081312.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P482814 AK006456.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P482814 NM_001081312.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P482814 AK006456.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P482814 NM_001081312.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P482814 AK006456.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P482814 NM_001081312.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P482814 AK006456.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23255003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23255008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23254936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23255016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23254988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23254935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23254954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23255021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23254990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23254965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23254995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23254941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23255042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23254985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23255036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23255013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23254938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23255024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23255005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23255043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23254943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23254939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23255007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23255018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23254949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23254952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23255001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23255020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23255002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23254960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23255039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23254955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23254947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23255040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23254945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23254937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23255025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23255011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23254999 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23255013 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23255008 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23255024 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03805398 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23255040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23254995 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23255020 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23255016 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03703714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03217837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23255005 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23254997 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23255033 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23254991 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23254992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23254985 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23255038 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23254994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23255017 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23254990 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23255003 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23255036 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23255021 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23255027 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23255018 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23255025 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03805397 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03425624 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23255028 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23254996 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23255041 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23255039 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23255030 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23255001 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03505722 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10516025 - -
          Affymetrix Mouse Gene 1.0 ST Array 10516026 - -
          Affymetrix HT MG-430 PM Array Plate 1440820_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10516024 - -
          Mouse Genome 430 2.0 Array 1440820_x_at - -
          Mouse Genome 430 2.0 Array 1440820_PM_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69469_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69469_at - -
          Affymetrix Mouse Clariom S Array PSR0400028043.mm.1 TC0400003453.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400028043.mm.1 NM_001081312.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400028043.mm.1 NM_001081312.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400028042.mm.1 TC0400003453.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400028042.mm.1 NM_001081312.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400028042.mm.1 NM_001081312.1 Refseq
          Mouse Expression Set 430 1440820_PM_x_at - -
          Mouse Expression Set 430 1440820_x_at - -
          Murine Genome U74v2 Set 167730_r_at - -
          Murine Genome U74v2 Set 167730_PM_r_at - -
          Affymetrix HT Mouse Genome 430B Array 1440820_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1440820_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 742371 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1258750 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1258750 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline