Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01192896_m1
          See other CK137956 GE Assays ›
          Gene Symbol:
          CK137956
          Gene Name:
          cDNA sequence CK137956
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 127927592 - 127988034 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01192896_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CK137956
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001134733.1 NP_001128205.1 6 - 7 1725 99
          GenBank mRNA BC147529.1 - 6 - 7 1725 9087439 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003572.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P280159 NM_001134733.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P280159 BC147529.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P280159 NM_001134733.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P280159 BC147529.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 959530 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 67260 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 74736 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 128566 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 804808 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 739339 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 865594 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 309340 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 653509 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 197083 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 799759 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 246507 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 578270 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 498903 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 439159 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 47556 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 362716 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 767302 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 771013 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 790230 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 620907 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 8167 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 464516 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 730707 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 936677 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 830543 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 351110 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 12901 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 42194 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 43522 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 109768 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P280159 NM_001134733.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P280159 BC147529.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P280159 NM_001134733.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P280159 BC147529.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P280159 NM_001134733.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P280159 BC147529.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P280159 NM_001134733.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P280159 BC147529.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10516497 - -
          Affymetrix Mouse Gene 1.0 ST Array 10516499 - -
          Affymetrix Mouse Gene 1.0 ST Array 10516500 - -
          Affymetrix Mouse Gene 1.0 ST Array 10516501 - -
          Affymetrix Mouse Gene 1.0 ST Array 10516496 - -
          Affymetrix Mouse Gene 1.0 ST Array 10516498 - -
          Affymetrix Mouse Gene 1.1 ST Array 10516495 - -
          Affymetrix Mouse Clariom S Array PSR0400029052.mm.1 TC0400003572.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400029052.mm.1 NM_001134733.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029052.mm.1 NM_001134733.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029051.mm.1 TC0400003572.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400029051.mm.1 NM_001134733.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029051.mm.1 NM_001134733.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 349907 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline