Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01197610_m1
          See other 1700008O03RIK GE Assays ›
          Gene Symbol:
          1700008O03Rik
          Gene Name:
          RIKEN cDNA 1700008O03 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 44358843 - 44385692 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01197610_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700008O03Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027049.1 NP_081325.1 5 - 6 574 88
          GenBank mRNA AK005784.1 - 5 - 6 575 88
          BC127243.1 - 3 - 4 690 88
          BC127244.1 - 3 - 4 690 88
          BF467486.1 - 1 - 2 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002958.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2182001 NM_027049.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2041410 BC127243.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2041410 BC127244.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2041410 BF467486.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2041410 AK005784.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2041410 NM_027049.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 153672 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 948359 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 457727 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 596140 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 829866 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 906484 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 781397 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 190301 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 935918 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 175662 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 45691 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101376 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142928 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2182001 NM_027049.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2041410 BC127243.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2041410 BC127244.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2041410 BF467486.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2041410 AK005784.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2041410 NM_027049.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142928 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01505880 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101376 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142928 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101376 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06202877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06124128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06349375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06079270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06241293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087081 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1454009_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10562767 - -
          Mouse Genome 430 2.0 Array 1454009_PM_at - -
          Mouse Genome 430 2.0 Array 1454009_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69349_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69349_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700026215.mm.1 TC0700002958.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700026215.mm.1 NM_027049.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026215.mm.1 NM_027049.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026216.mm.1 TC0700002958.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700026216.mm.1 NM_027049.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026216.mm.1 NM_027049.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026217.mm.1 TC0700002958.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700026217.mm.1 NM_027049.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026217.mm.1 NM_027049.1 Refseq
          Mouse Expression Set 430 1454009_at - -
          Mouse Expression Set 430 1454009_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1454009_at - -
          Affymetrix HT Mouse Genome 430B Array 1454009_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 869398 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 380500 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2603147 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2603147 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline