Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01199871_m1
          See other ARPC3 GE Assays ›
          Gene Symbol:
          Arpc3
          Gene Name:
          actin related protein 2/3 complex, subunit 3
          Gene Aliases:
          1110006A04Rik, 21kDa, AA408672, AI788639, p21-Ar, p21Arc
          Chromosome Location:
          Chr.5: 122391928 - 122406178 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01199871_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arpc3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019824.3 NP_062798.1 6 - 7 534 99
          GenBank mRNA AB038243.1 - 5 - 6 470 99
          AK003485.1 - 6 - 7 565 99
          AK005437.1 - 6 - 7 553 99
          AK171651.1 - 6 - 7 549 99
          BC013618.1 - 6 - 7 534 4013537 99
          BC054440.1 - 6 - 7 501 6395280 99
          BC055036.1 - 6 - 7 501 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001392.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Arpc3

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of actin filament polymerization
          Arp2/3 complex-mediated actin nucleation
          actin binding
          structural constituent of cytoskeleton
          actin filament binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24050077 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24050079 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24050081 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24050080 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2023952 AK171651.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2023952 BC013618.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2023952 NM_019824.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2023952 AK003485.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064792 AK005437.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064792 AK171651.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064792 BC054440.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064792 BC055036.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064792 AK003485.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2023953 BC013618.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2023953 NM_019824.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 252884 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 124155 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 939529 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 331484 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 835015 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 472417 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 196629 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 808367 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 104013 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158610 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153615 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2023953 BC013618.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2023953 NM_019824.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2023952 AK171651.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2023952 BC013618.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2023952 NM_019824.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2023952 AK003485.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2064792 AK005437.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2064792 AK171651.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2064792 BC054440.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2064792 BC055036.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2064792 AK003485.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P129767 AK005437.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P129767 BC013618.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P129767 BC054440.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P129767 BC055036.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P129767 NM_019824.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P686784 AK005437.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P686784 AK171651.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P686784 BC013618.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P686784 BC054440.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P686784 BC055036.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24050073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24050069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24050077 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24050076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24050084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24050088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24050086 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24050081 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24050080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24050079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24050074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24050070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24050071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24050082 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158610 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153615 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01207364 - -
          Whole Mouse Genome G4122A A_52_P686784 AK005437.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P686784 AK171651.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P686784 BC013618.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P686784 BC054440.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P686784 BC055036.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P129767 AK005437.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P129767 BC013618.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P129767 BC054440.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P129767 BC055036.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P129767 NM_019824.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110297 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153615 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158610 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04606127 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04190634 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04190633 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04606128 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04190636 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04606132 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24050081 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24050080 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24050079 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24050077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24050067 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24050081 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24050100 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04414766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04190562 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04545537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04190563 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24050102 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24050089 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24050068 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04606128 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24050093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04737455 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04190634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04737460 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24050094 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04499615 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04414769 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04606127 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04798170 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24050163 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04195206 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04798169 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24050087 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24050084 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24050086 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24050070 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04190633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24050082 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24050072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04203962 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04644145 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24050076 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10525402 - -
          Affymetrix Mouse Gene 1.0 ST Array 10525401 - -
          Affymetrix Mouse Gene 1.0 ST Array 10525398 - -
          Affymetrix Mouse Gene 1.0 ST Array 10525400 - -
          Affymetrix Mouse Gene 1.0 ST Array 10525399 - -
          Affymetrix Mouse Gene 1.0 ST Array 10525403 - -
          Affymetrix Mouse Gene 1.0 ST Array 10525405 - -
          Affymetrix Mouse Gene 1.0 ST Array 10525404 - -
          Affymetrix HT MG-430 PM Array Plate 1448279_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10525397 - -
          Mouse Genome 430 2.0 Array 1448279_at - -
          Mouse Genome 430 2.0 Array 1448279_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56378_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56378_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500012325.mm.1 TC0500001392.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500012325.mm.1 NM_019824.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500012327.mm.1 TC0500001392.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500012327.mm.1 NM_019824.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500012324.mm.1 TC0500001392.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500012324.mm.1 NM_019824.4 Refseq
          Mouse Expression Set 430 1448279_at - -
          Mouse Expression Set 430 1448279_PM_at - -
          Murine Genome U74v2 Set 95434_PM_at - -
          Murine Genome U74v2 Set 95434_at - -
          Mouse Genome 430A 2.0 Array 1448279_PM_at - -
          Mouse Genome 430A 2.0 Array 1448279_at - -
          Affymetrix HT Mouse Genome 430A Array 1448279_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448279_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 670441 - -
          Illumina mouse-6 v1.0 expression beadchip 670441 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000670441 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2603725 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2603725 - -
          mouse-6 v1.1 (Illumina) scl056378.6_20-S - -
          Illumina Sentrix mouse-6.v1 bead array 670441 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl056378.6_20-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 670441 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2603725 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2603725 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl056378.6_20-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline