Search
Search

Itgbl1| Gene Symbol: Itgbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145467.2 | NP_663442.2 | 7 - 8 | 1185 | 68 | |
| GenBank mRNA | AK029068.1 | - | 7 - 8 | 1152 | 68 | |
| AK047014.1 | - | 644 | 68 | |||
| AK051773.1 | - | 1223 | 68 | |||
| BC020152.1 | - | 7 - 8 | 1152 | 3708340 | 68 | |
Set Membership: |
|