Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01200871_m1
          See other PMS2 GE Assays ›
          Gene Symbol:
          Pms2
          Gene Name:
          postmeiotic segregation increased 2 (S. cerevisiae)
          Gene Aliases:
          AW555130, Pmsl2
          Chromosome Location:
          Chr.5: 143909974 - 143985719 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01200871_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pms2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008886.2 NP_032912.2 4 - 5 403 86
          GenBank mRNA AK017847.1 - 4 - 5 379 86
          AK034866.1 - 4 - 5 379 86
          AK083049.1 - 4 - 5 360 86
          AK146365.1 - 4 - 5 416 86
          BC022179.1 - 4 - 5 371 86
          BC058189.1 - 4 - 5 411 86
          BC066110.1 - 4 - 5 403 86
          BC083502.1 - 4 - 5 363 86
          BC086637.1 - 4 - 5 379 86
          BC110684.1 - 4 - 5 356 86
          BC139774.1 - 4 - 5 409 86
          BC141287.1 - 4 - 5 379 9055899 86

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the BC139774.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pms2

          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          mismatch repair
          cellular response to DNA damage stimulus
          meiotic nuclear division
          somatic hypermutation of immunoglobulin genes
          somatic recombination of immunoglobulin gene segments
          DNA binding
          single-stranded DNA binding
          nuclease activity
          endonuclease activity
          ATP binding
          hydrolase activity
          ATPase activity
          mismatched DNA binding
          single base insertion or deletion binding
          MutSalpha complex binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500016048.mm.1 TC0500003716.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500016048.mm.1 NM_008886.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016048.mm.1 NM_008886.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016049.mm.1 TC0500003716.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500016049.mm.1 NM_008886.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016049.mm.1 NM_008886.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016045.mm.1 TC0500003716.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500016045.mm.1 NM_008886.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016045.mm.1 NM_008886.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016044.mm.1 TC0500003716.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500016044.mm.1 NM_008886.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016044.mm.1 NM_008886.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 344558 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline