Search
Search

Fbxo21| Gene Symbol: Fbxo21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145564.3 | NP_663539.1 | 1 - 2 | 271 | 111 | |
| GenBank mRNA | AB093270.1 | - | 1 - 2 | 257 | 111 | |
| AK048131.1 | - | 1 - 2 | 250 | 111 | ||
| AK048542.1 | - | 1 - 2 | 250 | 111 | ||
| AK049723.1 | - | 1 - 2 | 272 | 111 | ||
| AK076521.1 | - | 1 - 2 | 268 | 111 | ||
| BC021871.1 | - | 1 - 2 | 308 | 3709413 | 111 | |
Set Membership: |
|