Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01209536_m1
          See other CYP3A25 GE Assays ›
          Gene Symbol:
          Cyp3a25
          Gene Name:
          cytochrome P450, family 3, subfamily a, polypeptide 25
          Gene Aliases:
          AI327008
          Chromosome Location:
          Chr.5: 145977194 - 146009639 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01209536_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp3a25
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019792.2 NP_062766.2 12 - 13 1491 63
          NR_030782.1 11 - 12 1403 63
          GenBank mRNA AF204959.1 - 12 - 13 1490 63
          AK149541.1 - 11 - 12 1404 63
          BC028855.1 - 12 - 13 1482 5124151 63
          Y11995.1 - 12 - 13 1493 63

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyp3a25

          Panther Classification:

          Molecular Function -

          oxygenase

          Gene Ontology Categories:

          Function(s) Process(es)

          oxidation-reduction process
          monooxygenase activity
          iron ion binding
          testosterone 16-alpha-hydroxylase activity
          oxidoreductase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced flavin or flavoprotein as one donor, and incorporation of one atom of oxygen
          heme binding
          metal ion binding
          testosterone 6-beta-hydroxylase activity
          aromatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2169259 Y11995.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2169259 AF204959.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2169259 AK149541.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2169259 BC028855.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2169259 NM_019792.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 644129 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 227501 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 69274 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 166646 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2169259 Y11995.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2169259 AF204959.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2169259 AK149541.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2169259 BC028855.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2169259 NM_019792.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P380314 Y11995.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P380314 AF204959.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P380314 AK149541.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P380314 BC028855.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P380314 NM_019792.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P489367 Y11995.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P489367 AF204959.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P489367 AK149541.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P489367 BC028855.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P489367 NM_019792.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24166592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24166583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24166767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24166581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24166765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24166761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24166745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24166575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24166762 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05780411 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01244345 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01244340 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01244346 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05780245 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05780225 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05780202 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05780435 - -
          Whole Mouse Genome G4122A A_52_P380314 Y11995.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P380314 AF204959.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P380314 AK149541.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P380314 BC028855.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P380314 NM_019792.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P489367 Y11995.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P489367 AF204959.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P489367 AK149541.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P489367 BC028855.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P489367 NM_019792.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107794 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05780368 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05780245 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04658032 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04658029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24166583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24166435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24166543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24166573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24166547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24166581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24166579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24166437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24166590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24166541 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166573 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166590 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166575 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166603 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166581 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166579 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04271295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166771 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166592 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10527497 - -
          Affymetrix Mouse Gene 1.0 ST Array 10527495 - -
          Affymetrix Mouse Gene 1.0 ST Array 10527501 - -
          Affymetrix Mouse Gene 1.0 ST Array 10527504 - -
          Affymetrix Mouse Gene 1.0 ST Array 10527506 - -
          Affymetrix Mouse Gene 1.0 ST Array 10527503 - -
          Affymetrix Mouse Gene 1.0 ST Array 10527496 - -
          Affymetrix Mouse Gene 1.0 ST Array 10527507 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535716 - -
          Affymetrix Mouse Gene 1.0 ST Array 10527502 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535717 - -
          Affymetrix Mouse Gene 1.0 ST Array 10527499 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535715 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535719 - -
          Affymetrix Mouse Gene 1.0 ST Array 10527498 - -
          Affymetrix Mouse Gene 1.0 ST Array 10527500 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535718 - -
          Affymetrix Mouse Gene 1.1 ST Array 10535714 - -
          Affymetrix Mouse Gene 1.1 ST Array 10527494 - -
          Mouse Genome 430 2.0 Array 1424973_at - -
          Mouse Genome 430 2.0 Array 1424973_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56388_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56388_at - -
          Affymetrix Mouse Clariom S Array PSR0500033065.mm.1 TC0500003601.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033065.mm.1 NM_019792.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033065.mm.1 NM_019792.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033065.mm.1 NR_030782.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033065.mm.1 NR_030782.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033064.mm.1 TC0500003601.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033064.mm.1 NM_019792.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033064.mm.1 NM_019792.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033064.mm.1 NR_030782.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033064.mm.1 NR_030782.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033066.mm.1 TC0500003601.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033066.mm.1 NM_019792.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033066.mm.1 NM_019792.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033066.mm.1 NR_030782.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033066.mm.1 NR_030782.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033067.mm.1 TC0500003601.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033067.mm.1 NM_019792.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033067.mm.1 NM_019792.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033067.mm.1 NR_030782.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033067.mm.1 NR_030782.1 Refseq
          Mouse Expression Set 430 1424973_PM_at - -
          Mouse Expression Set 430 1424973_at - -
          Murine Genome U74v2 Set 104024_PM_at - -
          Murine Genome U74v2 Set 104024_at - -
          Mouse Genome 430A 2.0 Array 1424973_PM_at - -
          Mouse Genome 430A 2.0 Array 1424973_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 506928 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1500348 - -
          MouseWG-6 v2 Expression BeadChip 6650427 - -
          Illumina mouse-6 v1.0 expression beadchip 1500348 - -
          Illumina mouse-6 v1.0 expression beadchip 6650427 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006650427 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001500348 - -
          mouse-6 v1.1 (Illumina) scl0056388.2_271-S - -
          mouse-6 v1.1 (Illumina) scl0056388.1_45-S - -
          Illumina Sentrix mouse-6.v1 bead array 1500348 - -
          Illumina Sentrix mouse-6.v1 bead array 6650427 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0056388.2_271-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0056388.1_45-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1500348 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6650427 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2928679 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2928679 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2740187 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2669354 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline