Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01212038_g1
          See other CYP2C66 GE Assays ›
          Gene Symbol:
          Cyp2c66
          Gene Name:
          cytochrome P450, family 2, subfamily c, polypeptide 66
          Gene Aliases:
          2010301M18Rik
          Chromosome Location:
          Chr.19: 39113898 - 39186756 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01212038_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp2c66
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011707.1 NP_001011707.1 2 - 3 364 60
          GenBank mRNA AY227734.1 - 2 - 3 364 60
          BC147819.1 - 2 - 3 361 9087536 60
          BC147824.1 - 2 - 3 361 9087541 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000564.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyp2c66

          Panther Classification:

          Molecular Function -

          oxidoreductase oxygenase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          hematopoietic progenitor cell differentiation
          xenobiotic metabolic process
          epoxygenase P450 pathway
          exogenous drug catabolic process
          oxidation-reduction process
          monooxygenase activity
          drug binding
          testosterone 16-alpha-hydroxylase activity
          arachidonic acid epoxygenase activity
          steroid hydroxylase activity
          oxidoreductase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced flavin or flavoprotein as one donor, and incorporation of one atom of oxygen
          oxygen binding
          heme binding
          caffeine oxidase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P471126 AK008688.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P471126 NM_028191.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P471126 AK008688.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P471126 NM_028191.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928026 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 870963 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 586616 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 577383 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 817718 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375746 BC035955.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375746 BC051169.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375746 NM_001081302.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375746 XM_001474968.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375746 XM_908972.3 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 89488 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 31057 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 195289 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 587968 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599799 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 849231 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 966343 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 813831 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 743818 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 237725 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 281977 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 6297 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 72398 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 111057 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 163500 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 155376 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 166325 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P471126 AK008688.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P471126 NM_028191.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P471126 AK008688.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P471126 NM_028191.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P471126 AK008688.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P471126 NM_028191.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P471126 AK008688.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P471126 NM_028191.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32104989 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32105004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32104996 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32104990 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32105005 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32104999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32104988 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32105000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32104986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32105001 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32104993 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32105007 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03595207 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09169055 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03595202 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09169078 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09169060 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09169094 - -
          Whole Mouse Genome G4122A A_51_P471126 AK008688.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P471126 NM_028191.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P471126 AK008688.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P471126 NM_028191.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09169094 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03595202 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09169063 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14323701 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14323697 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14323695 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14323704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32104982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32104995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32104983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32104998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32104986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32104988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32104996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32105001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32105000 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32104995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32105007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32104989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32104996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32104986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14404445 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32104988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32105001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32104984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32104993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14211641 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32104990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14323698 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32105003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32104998 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10463026 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463024 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463025 - -
          Affymetrix Mouse Gene 1.1 ST Array 10463023 - -
          Affymetrix Mouse Clariom S Array PSR1900005089.mm.1 NM_001011707.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005089.mm.1 NM_001011707.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005089.mm.1 TC1900000564.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900005092.mm.1 NM_001011707.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005092.mm.1 NM_001011707.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005092.mm.1 TC1900000564.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900005094.mm.1 NM_001011707.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005094.mm.1 NM_001011707.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005094.mm.1 TC1900000564.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900005095.mm.1 NM_001011707.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005095.mm.1 NM_001011707.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005095.mm.1 TC1900000564.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900005096.mm.1 NM_001011707.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005096.mm.1 NM_001011707.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005096.mm.1 TC1900000564.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900005087.mm.1 NM_001011707.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005087.mm.1 NM_001011707.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005087.mm.1 TC1900000564.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900005090.mm.1 NM_001011707.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005090.mm.1 NM_001011707.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005090.mm.1 TC1900000564.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900005093.mm.1 NM_001011707.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005093.mm.1 NM_001011707.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005093.mm.1 TC1900000564.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900005088.mm.1 NM_001011707.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005088.mm.1 NM_001011707.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005088.mm.1 TC1900000564.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900005091.mm.1 NM_001011707.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005091.mm.1 NM_001011707.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005091.mm.1 TC1900000564.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2765099 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1241162 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2765099 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1241162 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline