Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01216737_m1
          See other SAC3D1 GE Assays ›
          Gene Symbol:
          Sac3d1
          Gene Name:
          SAC3 domain containing 1
          Gene Aliases:
          2410004C24Rik, AU015683, shd1
          Chromosome Location:
          Chr.19: 6116004 - 6118586 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01216737_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sac3d1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133678.3 NP_598439.3 1 - 2 790 75
          GenBank mRNA AK011298.1 - 1 - 2 702 75
          AK014138.1 - 1 - 2 744 75
          AK075930.1 - 1 - 2 750 75
          BC144812.1 - 1 - 2 854 9053360 75
          BC145789.1 - 1 - 2 854 40131092 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900001002.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sac3d1

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cell cycle
          mitotic nuclear division
          negative regulation of JAK-STAT cascade
          regulation of immune response
          spindle assembly
          centrosome duplication
          cell division
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31932822 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31932825 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31932820 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31932826 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31932824 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31932823 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31932821 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2168628 BC145789.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2168628 BC144812.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2168628 AK011298.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2168628 AK014138.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2168628 AJ131957.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2083023 AK011298.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2083023 AK014138.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2083023 AJ131957.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2083023 AK075930.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2083023 AK139556.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 535756 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 252562 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119958 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2168628 BC145789.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2168628 BC144812.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2168628 AK011298.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2168628 AK014138.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2168628 AJ131957.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2083023 AK011298.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2083023 AK014138.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2083023 AJ131957.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2083023 AK075930.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2083023 AK139556.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P162624 BC145789.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P162624 BC144812.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P162624 AK011298.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P162624 AK014138.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P162624 AJ131957.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932808 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932817 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932813 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932812 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932821 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932823 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932810 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932819 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932820 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932818 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932827 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932816 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P119958 - -
          Whole Mouse Genome G4122A A_51_P162624 BC145789.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P162624 BC144812.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P162624 AK011298.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P162624 AK014138.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P162624 AJ131957.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114265 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P119958 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14470998 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14470991 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31932823 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31932824 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31932826 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31932825 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31932821 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31932820 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31932822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14470991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932813 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14470998 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14470987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14470999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932808 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10465375 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465376 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465378 - -
          Affymetrix HT MG-430 PM Array Plate 1417036_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1417035_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1456608_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10465374 - -
          Mouse Genome 430 2.0 Array 1417035_PM_at - -
          Mouse Genome 430 2.0 Array 1417035_at - -
          Mouse Genome 430 2.0 Array 1417036_at - -
          Mouse Genome 430 2.0 Array 1417036_PM_at - -
          Mouse Genome 430 2.0 Array 1456608_at - -
          Mouse Genome 430 2.0 Array 1456608_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66406_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66406_at - -
          Affymetrix Mouse Clariom S Array PSR1900009253.mm.1 TC1900001002.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009251.mm.1 NM_133678.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009251.mm.1 NM_133678.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009251.mm.1 TC1900001002.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009252.mm.1 NM_133678.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009252.mm.1 NM_133678.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009252.mm.1 TC1900001002.mm.1 MTA TC
          Mouse Expression Set 430 1417036_PM_at - -
          Mouse Expression Set 430 1456608_PM_at - -
          Mouse Expression Set 430 1456608_at - -
          Mouse Expression Set 430 1417035_at - -
          Mouse Expression Set 430 1417036_at - -
          Mouse Expression Set 430 1417035_PM_at - -
          Murine Genome U74v2 Set 162131_PM_f_at - -
          Murine Genome U74v2 Set 97891_at - -
          Murine Genome U74v2 Set 161135_PM_f_at - -
          Murine Genome U74v2 Set 162131_f_at - -
          Murine Genome U74v2 Set 97891_PM_at - -
          Murine Genome U74v2 Set 161135_f_at - -
          Murine Genome U74v2 Set 169331_at - -
          Murine Genome U74v2 Set 169358_i_at - -
          Murine Genome U74v2 Set 169331_PM_at - -
          Murine Genome U74v2 Set 169358_PM_i_at - -
          Mouse Genome 430A 2.0 Array 1417035_at - -
          Mouse Genome 430A 2.0 Array 1456608_at - -
          Mouse Genome 430A 2.0 Array 1417035_PM_at - -
          Mouse Genome 430A 2.0 Array 1417036_at - -
          Mouse Genome 430A 2.0 Array 1417036_PM_at - -
          Mouse Genome 430A 2.0 Array 1456608_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417036_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1456608_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417036_at - -
          Affymetrix HT Mouse Genome 430A Array 1417035_at - -
          Affymetrix HT Mouse Genome 430A Array 1417035_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1456608_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 923933 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1770114 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001770114 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2691706 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2691706 - -
          mouse-6 v1.1 (Illumina) scl53527.2.1_48-S - -
          Illumina Sentrix mouse-6.v1 bead array 1770114 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl53527.2.1_48-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1770114 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2691706 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2691706 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline