Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01217231_g1
          See other TMEM129 GE Assays ›
          Gene Symbol:
          Tmem129
          Gene Name:
          transmembrane protein 129
          Gene Aliases:
          0610039G24Rik, 2410015J15Rik, AA409575
          Chromosome Location:
          Chr.5: 33653216 - 33657911 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01217231_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem129
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026698.2 NP_080974.2 3 - 4 845 68
          GenBank mRNA AK002836.1 - 3 - 4 894 68
          AK172044.1 - 3 - 4 974 68
          BC029088.1 - 3 - 4 891 3587662 68
          BC094042.1 - 3 - 4 845 5098723 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002221.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem129

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          response to unfolded protein
          ER-associated ubiquitin-dependent protein catabolic process
          retrograde protein transport, ER to cytosol
          UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
          ligase activity
          ribosomal S6-glutamic acid ligase activity
          coenzyme F420-0 gamma-glutamyl ligase activity
          coenzyme F420-2 alpha-glutamyl ligase activity
          metal ion binding
          ubiquitin protein ligase activity
          protein-glycine ligase activity
          protein-glycine ligase activity, initiating
          protein-glycine ligase activity, elongating
          tubulin-glycine ligase activity
          protein-glutamic acid ligase activity
          tubulin-glutamic acid ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23590346 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23590340 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23590342 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23590338 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23590343 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23590344 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23590341 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23590345 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23590347 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P101982 BC094042.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P101982 AK010497.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P101982 AK172044.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P101982 NM_026698.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P101982 BC094042.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P101982 AK010497.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P101982 AK172044.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P101982 NM_026698.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 612518 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 622037 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 593395 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100206 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P101982 BC094042.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P101982 AK010497.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P101982 AK172044.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P101982 NM_026698.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P101982 BC094042.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P101982 AK010497.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P101982 AK172044.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P101982 NM_026698.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23590303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23590310 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23590311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23590307 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23590336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23590338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23590322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23590309 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23590308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23590330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23590327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23590335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23590315 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23590313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23590320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23590342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23590306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23590324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23590325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23590316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23590346 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23590305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23590344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23590332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23590337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23590317 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100206 - -
          Whole Mouse Genome G4122A A_52_P101982 BC094042.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P101982 AK010497.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P101982 AK172044.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P101982 NM_026698.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P101982 BC094042.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P101982 AK010497.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P101982 AK172044.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P101982 NM_026698.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468037 BC094042.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468037 AK172044.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468037 BC029088.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468037 NM_026698.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468037 AK002836.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100206 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04544684 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04611013 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23590338 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23590346 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23590341 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23590347 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23590343 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23590342 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23590340 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23590345 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23590344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23590325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23590332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04611015 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04611009 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23590327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23590330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23590325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04611032 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04611025 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04611030 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23590324 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23590335 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23590322 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04611028 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04611042 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23590303 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23590328 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04611040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04544678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04611022 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04346551 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04611020 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04544703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23590340 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10529308 - -
          Affymetrix Mouse Gene 1.0 ST Array 10529306 - -
          Affymetrix Mouse Gene 1.0 ST Array 10529307 - -
          Affymetrix Mouse Gene 1.0 ST Array 10529309 - -
          Affymetrix Mouse Gene 1.0 ST Array 10529310 - -
          Affymetrix HT MG-430 PM Array Plate 1415715_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1431015_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10529305 - -
          Mouse Genome 430 2.0 Array 1415715_at - -
          Mouse Genome 430 2.0 Array 1415715_PM_at - -
          Mouse Genome 430 2.0 Array 1431015_PM_at - -
          Mouse Genome 430 2.0 Array 1431015_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68366_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68366_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500020297.mm.1 TC0500002221.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500020297.mm.1 NM_026698.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020297.mm.1 NM_026698.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020298.mm.1 TC0500002221.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500020298.mm.1 NM_026698.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020298.mm.1 NM_026698.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020299.mm.1 TC0500002221.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500020299.mm.1 NM_026698.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020299.mm.1 NM_026698.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020295.mm.1 TC0500002221.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500020295.mm.1 NM_026698.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020295.mm.1 NM_026698.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020296.mm.1 TC0500002221.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500020296.mm.1 NM_026698.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020296.mm.1 NM_026698.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020302.mm.1 TC0500002221.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500020302.mm.1 NM_026698.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020302.mm.1 NM_026698.2 Refseq
          Mouse Expression Set 430 1431015_at - -
          Mouse Expression Set 430 1431015_PM_at - -
          Murine Genome U74v2 Set 161542_r_at - -
          Murine Genome U74v2 Set 161542_PM_r_at - -
          Affymetrix HT Mouse Genome 430A Array 1415715_at - -
          Affymetrix HT Mouse Genome 430A Array 1415715_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431015_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1415715_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431015_at - -
          Affymetrix HT Mouse Genome 430B Array 1415715_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 460371 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2688871 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2429215 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2688871 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline