Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01217835_m1
          See other CGREF1 GE Assays ›
          Gene Symbol:
          Cgref1
          Gene Name:
          cell growth regulator with EF hand domain 1
          Gene Aliases:
          1110004G24Rik, AW045201, Cgr11
          Chromosome Location:
          Chr.5: 30933143 - 30945480 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01217835_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cgref1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001160149.1 NP_001153621.1 2 - 3 140 61
          NM_026770.1 NP_081046.1 2 - 3 224 61
          GenBank mRNA AK003420.1 - 2 - 3 151 61
          BC023116.1 - 2 - 3 224 4206019 61
          BQ555687.1 - 2 - 3 340 61
          CB202031.1 - 2 - 3 141 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002168.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cell cycle
          cell cycle arrest
          cell adhesion
          negative regulation of cell growth
          molecular_function
          calcium ion binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P372550 BQ043218.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P372550 NM_001160149.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P372550 NM_026770.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P372550 AK003420.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P372550 BC023116.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P372550 BQ043218.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P372550 NM_001160149.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P372550 NM_026770.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P372550 AK003420.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P372550 BC023116.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 87025 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 810458 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 769611 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 207263 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 204886 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 798359 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 430863 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 97213 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 43454 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158640 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139981 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P372550 BQ043218.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P372550 NM_001160149.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P372550 NM_026770.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P372550 AK003420.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P372550 BC023116.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P372550 BQ043218.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P372550 NM_001160149.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P372550 NM_026770.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P372550 AK003420.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P372550 BC023116.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P372550 BQ043218.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P372550 NM_001160149.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P372550 NM_026770.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P372550 AK003420.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P372550 BC023116.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P372550 BQ043218.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P372550 NM_001160149.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P372550 NM_026770.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P372550 AK003420.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P372550 BC023116.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158640 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01065584 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139981 - -
          Whole Mouse Genome G4122A A_51_P372550 BQ043218.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P372550 NM_001160149.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P372550 NM_026770.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P372550 AK003420.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P372550 BC023116.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P372550 BQ043218.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P372550 NM_001160149.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P372550 NM_026770.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P372550 AK003420.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P372550 BC023116.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139981 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158640 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10529036 - -
          Affymetrix Mouse Gene 1.0 ST Array 10529038 - -
          Affymetrix Mouse Gene 1.0 ST Array 10529037 - -
          Affymetrix Mouse Gene 1.0 ST Array 10529035 - -
          Affymetrix Mouse Gene 1.0 ST Array 10529040 - -
          Affymetrix Mouse Gene 1.0 ST Array 10529039 - -
          Affymetrix HT MG-430 PM Array Plate 1424529_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1424528_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10529034 - -
          Mouse Genome 430 2.0 Array 1424529_PM_s_at - -
          Mouse Genome 430 2.0 Array 1424528_PM_at - -
          Mouse Genome 430 2.0 Array 1424529_s_at - -
          Mouse Genome 430 2.0 Array 1424528_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68567_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68567_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500019677.mm.1 TC0500002168.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019677.mm.1 NM_001160149.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019677.mm.1 NM_001160149.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019677.mm.1 NM_026770.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019677.mm.1 NM_026770.1 Refseq
          Mouse Expression Set 430 1424528_at - -
          Mouse Expression Set 430 1424529_s_at - -
          Mouse Expression Set 430 1424528_PM_at - -
          Mouse Expression Set 430 1424529_PM_s_at - -
          Murine Genome U74v2 Set 166986_PM_r_at - -
          Murine Genome U74v2 Set 166986_r_at - -
          Mouse Genome 430A 2.0 Array 1424528_at - -
          Mouse Genome 430A 2.0 Array 1424528_PM_at - -
          Mouse Genome 430A 2.0 Array 1424529_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1424529_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1424529_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1424529_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1424528_at - -
          Affymetrix HT Mouse Genome 430A Array 1424528_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 555816 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4540546 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2601386 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2601386 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline