Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01217843_m1
          See other CYTL1 GE Assays ›
          Gene Symbol:
          Cytl1
          Gene Name:
          cytokine-like 1
          Gene Aliases:
          4930443F05Rik, 4Cytl1, C17, Cyt1, Gm147
          Chromosome Location:
          Chr.5: 37735519 - 37739820 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01217843_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cytl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081106.1 NP_001074575.1 3 - 4 367 70
          GenBank mRNA AK076770.1 - 2 - 3 259 70
          BC063103.1 - 3 - 4 327 70
          BC116224.1 - 3 - 4 40091368 70
          BC116225.1 - 3 - 4 40091369 70
          EF108311.1 - 3 - 4 338 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000425.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          cytokine intercellular signal molecule

          Gene Ontology Categories:

          Function(s)

          chondrocyte differentiation
          positive regulation of transcription from RNA polymerase II promoter
          inner ear development
          chondroitin sulfate proteoglycan biosynthetic process
          positive regulation of sequence-specific DNA binding transcription factor activity
          cartilage homeostasis

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P631547 NM_001081106.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P631547 AK076770.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P631547 BC063103.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P631547 NM_001081106.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P631547 AK076770.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P631547 BC063103.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 287770 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 589966 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 309858 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 65950 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P110395 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P631547 NM_001081106.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P631547 AK076770.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P631547 BC063103.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P631547 NM_001081106.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P631547 AK076770.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P631547 BC063103.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P110395 - -
          Whole Mouse Genome G4122A A_52_P631547 NM_001081106.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P631547 AK076770.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P631547 BC063103.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P631547 NM_001081106.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P631547 AK076770.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P631547 BC063103.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P110395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23615189 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615156 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615212 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04628420 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04920396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04203902 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615237 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615239 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04275685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615199 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04203881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04171808 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615201 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615185 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04628430 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04960686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04920392 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615157 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615150 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615230 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04379595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04960690 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04960684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615249 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04403394 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615155 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04628427 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615172 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04403388 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615194 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615241 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615242 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04628418 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615177 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04203893 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615189 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04203900 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615198 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615205 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615245 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615183 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04652992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615232 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615149 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04275686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04582920 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615217 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615168 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04633205 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04421673 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04364637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615256 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04960687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615228 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615169 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615196 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615153 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615203 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615192 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04275680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615160 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615178 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04920400 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04628426 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04920395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23615146 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10521540 - -
          Affymetrix Mouse Gene 1.0 ST Array 10521542 - -
          Affymetrix Mouse Gene 1.0 ST Array 10521539 - -
          Affymetrix Mouse Gene 1.0 ST Array 10521541 - -
          Affymetrix Mouse Gene 1.0 ST Array 10521538 - -
          Affymetrix HT MG-430 PM Array Plate 1456793_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10521537 - -
          Mouse Genome 430 2.0 Array 1456793_PM_at - -
          Mouse Genome 430 2.0 Array 1456793_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 231162_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 231162_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500004243.mm.1 TC0500000425.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500004243.mm.1 NM_001081106.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500004243.mm.1 NM_001081106.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500004244.mm.1 TC0500000425.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500004244.mm.1 NM_001081106.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500004244.mm.1 NM_001081106.1 Refseq
          Mouse Expression Set 430 1456793_at - -
          Mouse Expression Set 430 1456793_PM_at - -
          Murine Genome U74v2 Set 170350_PM_at - -
          Murine Genome U74v2 Set 168183_PM_f_at - -
          Murine Genome U74v2 Set 168183_f_at - -
          Murine Genome U74v2 Set 170350_at - -
          Affymetrix HT Mouse Genome 430B Array 1456793_at - -
          Affymetrix HT Mouse Genome 430B Array 1456793_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 347591 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1221146 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1221146 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2877029 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2877029 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline