Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01218177_m1
          See other JTB GE Assays ›
          Gene Symbol:
          Jtb
          Gene Name:
          jumping translocation breakpoint
          Gene Aliases:
          Gm622
          Chromosome Location:
          Chr.3: 90231597 - 90235840 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01218177_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Jtb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_206924.2 NP_996807.1 3 - 4 566 102
          GenBank mRNA AB016490.1 - 3 - 4 511 102
          AK004469.1 - 3 - 4 478 102
          AK089048.1 - 3 - 4 478 102
          AK146337.1 - 3 - 4 567 102
          AK215601.1 - 1 - 2 80 102
          BC008139.1 - 3 - 4 530 3582930 102
          BG077727.2 - 2 - 3 132 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300000784.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Jtb

          Gene Ontology Categories:

          Function(s) Process(es)

          cytokinesis
          apoptotic process
          cell cycle
          mitotic nuclear division
          apoptotic mitochondrial changes
          regulation of cell proliferation
          positive regulation of protein kinase activity
          cell division
          protein kinase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22324051 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22324050 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22324048 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22324052 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22324049 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P603976 AK146337.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P603976 AK215601.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P603976 BC008139.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P603976 BG077727.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P603976 AK089048.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P603976 AK146337.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P603976 AK215601.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P603976 BC008139.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P603976 BG077727.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P603976 AK089048.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 25335 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 236873 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 823172 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 55749 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107512 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P603976 AK146337.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P603976 AK215601.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P603976 BC008139.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P603976 BG077727.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P603976 AK089048.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P603976 AK146337.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P603976 AK215601.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P603976 BC008139.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P603976 BG077727.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P603976 AK089048.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324046 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324035 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107512 - -
          Whole Mouse Genome G4122A A_51_P209135 AB016490.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P603976 AK146337.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P603976 AK215601.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P603976 BC008139.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P603976 BG077727.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P603976 AK089048.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P603976 AK146337.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P603976 AK215601.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P603976 BC008139.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P603976 BG077727.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P603976 AK089048.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138456 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107512 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02624617 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02624607 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02624611 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02624622 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22324050 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22324048 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22324052 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22324049 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22324051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324123 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324031 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324060 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324065 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324033 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324028 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324025 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324042 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324061 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324066 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02624610 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324036 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324038 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324069 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02624611 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324032 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324030 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02650440 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02624607 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02393134 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324056 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324068 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02935446 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02624614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324044 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324037 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324063 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02624609 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03004073 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10493704 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493708 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493705 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493707 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493706 - -
          Affymetrix HT MG-430 PM Array Plate 1424283_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10493703 - -
          Mouse Genome 430 2.0 Array 1424283_PM_at - -
          Mouse Genome 430 2.0 Array 1424283_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 23922_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 23922_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0300006086.mm.1 NM_206924.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006086.mm.1 NM_206924.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006086.mm.1 TC0300000784.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006089.mm.1 NM_206924.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006089.mm.1 NM_206924.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006089.mm.1 TC0300000784.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006090.mm.1 NM_206924.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006090.mm.1 NM_206924.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006090.mm.1 TC0300000784.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006091.mm.1 NM_206924.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006091.mm.1 NM_206924.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006091.mm.1 TC0300000784.mm.1 MTA TC
          Mouse Expression Set 430 1424283_PM_at - -
          Mouse Expression Set 430 1424283_at - -
          Murine Genome U74v2 Set 102790_at - -
          Murine Genome U74v2 Set 102790_PM_at - -
          Mouse Genome 430A 2.0 Array 1424283_PM_at - -
          Mouse Genome 430A 2.0 Array 1424283_at - -
          Affymetrix HT Mouse Genome 430A Array 1424283_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424283_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 362025 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6980400 - -
          Illumina mouse-6 v1.0 expression beadchip 6980400 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006980400 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1226138 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1226138 - -
          mouse-6 v1.1 (Illumina) scl22956.4.1_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 6980400 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl22956.4.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6980400 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1226138 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1226138 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl22956.4.1_1-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline