Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01218907_m1
          See other A430093F15RIK GE Assays ›
          Gene Symbol:
          A430093F15Rik
          Gene Name:
          RIKEN cDNA A430093F15 gene
          Gene Aliases:
          Gm674
          Chromosome Location:
          Chr.19: 10740947 - 10786043 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01218907_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A430093F15Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027805.1 2 - 3 1375 71
          GenBank mRNA AK079853.1 - 2 - 3 1379 71
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA NM_207529.1 - 1379 Chr. - - [-] on Build 71

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK079853.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene A430093F15Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2019503 AK079853.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2019503 NR_027805.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 459890 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 557891 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 163043 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 339401 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 447246 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 48825 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 762398 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 769828 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 260819 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 356836 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 741663 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 881783 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 134082 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 384339 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 187314 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 223839 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 342413 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 701855 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 577480 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 759711 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 273092 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 490647 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 771304 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 741704 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 676168 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 148669 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 389380 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 21537 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 664603 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 167866 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 110911 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 135614 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 17027 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 146093 - -
          Whole Mouse Genome G4122A A_52_P526740 AK079853.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P526740 BC064125.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P526740 NR_027805.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P14486622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952682 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952663 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952747 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952751 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14465090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14398623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952741 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14429159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14444332 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952740 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14274834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952742 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14183643 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14429155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952666 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952746 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14289742 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952749 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31952745 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10461554 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461557 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461556 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461555 - -
          Affymetrix HT MG-430 PM Array Plate 1444299_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10461553 - -
          Mouse Genome 430 2.0 Array 1444299_PM_at - -
          Mouse Genome 430 2.0 Array 1444299_at - -
          Affymetrix Mouse Clariom S Array PSR1900002630.mm.1 NR_027805.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002630.mm.1 NR_027805.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002630.mm.1 TC1900000209.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002632.mm.1 NR_027805.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002632.mm.1 NR_027805.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002632.mm.1 TC1900000209.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002631.mm.1 NR_027805.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002631.mm.1 NR_027805.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002631.mm.1 TC1900000209.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002633.mm.1 NR_027805.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002633.mm.1 NR_027805.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002633.mm.1 TC1900000209.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002629.mm.1 NR_027805.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002629.mm.1 NR_027805.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002629.mm.1 TC1900000209.mm.1 MTA TC
          Mouse Expression Set 430 1444299_at - -
          Mouse Expression Set 430 1444299_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1444299_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1444299_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 421942 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5900670 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline