Search
Search
| Gene Symbol: Coq3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172687.1 | NP_766275.1 | 4 - 5 | 505 | 69 | |
| GenBank mRNA | AK028680.1 | - | 4 - 5 | 505 | 69 | |
| AK035318.1 | - | 4 - 5 | 500 | 69 | ||
| AK142287.1 | - | 4 - 5 | 443 | 69 | ||
| AK166475.1 | - | 4 - 5 | 460 | 69 | ||
| AK170488.1 | - | 4 - 5 | 500 | 69 | ||
| BC060960.1 | - | 4 - 5 | 507 | 30312962 | 69 | |
Set Membership: |
|