Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01221714_g1
          See other PRR14L GE Assays ›
          Gene Symbol:
          Prr14l
          Gene Name:
          proline rich 14-like
          Gene Aliases:
          6030436E02Rik, C330019G07Rik, Prl14l
          Chromosome Location:
          Chr.5: 32789206 - 32854338 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01221714_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prr14l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_194340.2 NP_919321.2 9 - 10 5843 107
          GenBank mRNA AK049285.1 - 2 - 3 175 107
          AY372183.1 - 4 - 5 1064 107
          BC059908.1 - 2 - 3 275 107
          BC094374.1 - 4 - 5 1390 30531815 107

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prr14l

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500020134.mm.1 TC0500003726.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500020134.mm.1 NM_194340.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020134.mm.1 NM_194340.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020133.mm.1 TC0500003726.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500020133.mm.1 NM_194340.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020133.mm.1 NM_194340.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020135.mm.1 TC0500003726.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500020135.mm.1 NM_194340.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020135.mm.1 NM_194340.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020130.mm.1 TC0500003726.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500020130.mm.1 NM_194340.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020130.mm.1 NM_194340.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020136.mm.1 TC0500003726.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500020136.mm.1 NM_194340.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020136.mm.1 NM_194340.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020132.mm.1 TC0500003726.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500020132.mm.1 NM_194340.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020132.mm.1 NM_194340.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline