Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01224565_m1
          See other DOCK9 GE Assays ›
          Gene Symbol:
          Dock9
          Gene Name:
          dedicator of cytokinesis 9
          Gene Aliases:
          AA959601, AW538057, B230309H04Rik, D14Wsu89e, Zizimin1, mKIAA1058
          Chromosome Location:
          Chr.14: 121542039 - 121797897 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01224565_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dock9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081039.1 NP_001074508.1 32 - 33 3669 69
          NM_001128307.1 NP_001121779.1 32 - 33 4074 69
          NM_001128308.1 NP_001121780.1 32 - 33 4074 69
          NM_134074.2 NP_598835.2 32 - 33 3690 69
          GenBank mRNA AK122431.1 - 32 - 33 4073 69
          BC046250.1 - 26 - 27 2976 69
          BC052947.1 - 26 - 27 2988 69
          BC157964.1 - 32 - 33 3614 9007289 69
          BC158050.1 - 32 - 33 4005 9007354 69
          BC158127.1 - 32 - 33 3573 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400002740.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dock9

          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          small GTPase mediated signal transduction
          positive regulation of GTPase activity
          guanyl-nucleotide exchange factor activity
          Rho guanyl-nucleotide exchange factor activity
          Rho GTPase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1400020421.mm.1 NM_001081039.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400020421.mm.1 NM_001081039.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400020421.mm.1 NM_001128307.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400020421.mm.1 NM_001128307.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400020421.mm.1 NM_001128308.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400020421.mm.1 NM_001128308.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400020421.mm.1 NM_134074.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400020421.mm.1 NM_134074.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400020421.mm.1 TC1400002740.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400020420.mm.1 NM_001081039.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400020420.mm.1 NM_001081039.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400020420.mm.1 NM_001128307.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400020420.mm.1 NM_001128307.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400020420.mm.1 NM_001128308.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400020420.mm.1 NM_001128308.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400020420.mm.1 NM_134074.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400020420.mm.1 NM_134074.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400020420.mm.1 TC1400002740.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline