Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01224995_m1
          See other 1700031F05RIK GE Assays ›
          Gene Symbol:
          1700031F05Rik
          Gene Name:
          RIKEN cDNA 1700031F05 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 102859186 - 102867024 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01224995_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700031F05Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028496.1 NP_082772.1 6 - 7 787 57
          GenBank mRNA AK006578.1 - 6 - 7 788 57

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P269020 NM_028496.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P269020 AK006578.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P269020 NM_028496.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P269020 AK006578.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P269020 NM_028496.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P269020 AK006578.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P269020 NM_028496.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P269020 AK006578.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P32570782 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32570805 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15210992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15119615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15119602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32570765 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432116_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10606146 - -
          Mouse Genome 430 2.0 Array 1432116_PM_at - -
          Mouse Genome 430 2.0 Array 1432116_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73300_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73300_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00017399.mm.1 NM_028496.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017399.mm.1 NM_028496.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017399.mm.1 TC0X00002760.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017395.mm.1 NM_028496.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017395.mm.1 NM_028496.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017395.mm.1 TC0X00002760.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017397.mm.1 NM_028496.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017397.mm.1 NM_028496.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017397.mm.1 TC0X00002760.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017401.mm.1 NM_028496.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017401.mm.1 NM_028496.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017401.mm.1 TC0X00002760.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017400.mm.1 NM_028496.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017400.mm.1 NM_028496.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017400.mm.1 TC0X00002760.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017403.mm.1 NM_028496.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017403.mm.1 NM_028496.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017403.mm.1 TC0X00002760.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017398.mm.1 NM_028496.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017398.mm.1 NM_028496.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017398.mm.1 TC0X00002760.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017402.mm.1 NM_028496.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017402.mm.1 NM_028496.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017402.mm.1 TC0X00002760.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017396.mm.1 NM_028496.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017396.mm.1 NM_028496.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017396.mm.1 TC0X00002760.mm.1 MTA TC
          Mouse Expression Set 430 1432116_PM_at - -
          Mouse Expression Set 430 1432116_at - -
          Affymetrix HT Mouse Genome 430B Array 1432116_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432116_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 625798 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102120128 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1243172 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1244621 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline