Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01233548_m1
          See other GAL3ST3 GE Assays ›
          Gene Symbol:
          Gal3st3
          Gene Name:
          galactose-3-O-sulfotransferase 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 5298331 - 5308739 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01233548_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gal3st3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024717.2 NP_001019888.1 2 - 3 402 69
          GenBank mRNA BC066996.1 - 2 - 3 392 5707267 69
          BU612752.1 - 2 - 3 402 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000060.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          biosynthetic process
          N-acetylglucosamine 6-O-sulfotransferase activity
          N-acetylgalactosamine 4-O-sulfotransferase activity
          galactosylceramide sulfotransferase activity
          heparan sulfate 2-O-sulfotransferase activity
          HNK-1 sulfotransferase activity
          transferase activity
          heparan sulfate 6-O-sulfotransferase activity
          trans-9R,10R-dihydrodiolphenanthrene sulfotransferase activity
          1-phenanthrol sulfotransferase activity
          3-phenanthrol sulfotransferase activity
          4-phenanthrol sulfotransferase activity
          trans-3,4-dihydrodiolphenanthrene sulfotransferase activity
          9-phenanthrol sulfotransferase activity
          2-phenanthrol sulfotransferase activity
          phenanthrol sulfotransferase activity
          1-hydroxypyrene sulfotransferase activity
          galactose 3-O-sulfotransferase activity
          proteoglycan sulfotransferase activity
          cholesterol sulfotransferase activity
          hydroxyjasmonate sulfotransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31927971 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31927961 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31927968 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31927962 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31927972 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31927969 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31927966 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31927970 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2125401 BC066996.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2125401 NM_001024717.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 306032 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 411327 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 960573 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 224345 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 783683 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 54725 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104580 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137135 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2125401 BC066996.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2125401 NM_001024717.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31927917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31927924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31927927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31927922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31927920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31927914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31927929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31927916 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31927919 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31927915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31927918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31927909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31927912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31927923 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31927926 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03542729 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104580 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137135 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104580 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137135 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14196117 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14196112 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31927971 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31927966 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31927968 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31927961 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31927970 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31927962 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31927969 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31927972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31927966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31927949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14503102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31927947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31927941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14196124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31927927 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31927897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31927953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31927931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14143810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31927957 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31927907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31927912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14196122 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31927914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31927933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31927935 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14503096 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31927928 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31927942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31927901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31927924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31927946 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31927902 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31927943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31927898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31927944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31927909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31927904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31927926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31927918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31927939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14399371 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31927937 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31927906 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10460557 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460559 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460558 - -
          Affymetrix Mouse Gene 1.1 ST Array 10460556 - -
          Affymetrix Mouse Clariom S Array PSR1900000749.mm.1 NM_001024717.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000749.mm.1 NM_001024717.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000749.mm.1 TC1900000060.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000748.mm.1 NM_001024717.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000748.mm.1 NM_001024717.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000748.mm.1 TC1900000060.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000746.mm.1 NM_001024717.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000746.mm.1 NM_001024717.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000746.mm.1 TC1900000060.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 472000 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1224213 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3106849 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3033192 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3106849 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3033192 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline