Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01234249_g1
          See other MIEF2 GE Assays ›
          Gene Symbol:
          Mief2
          Gene Name:
          mitochondrial elongation factor 2
          Gene Aliases:
          AI482195, Gm11, Smcr7
          Chromosome Location:
          Chr.11: 60728398 - 60732951 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01234249_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mief2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009927.2 NP_001009927.1 3 - 4 485 76
          GenBank mRNA AK156917.1 - 3 - 4 468 76
          AK171444.1 - 2 - 3 610 76
          AK211527.1 - 2 - 3 77
          BC088984.1 - 3 - 4 374 5708020 76

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK171444.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mief2

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrion organization
          positive regulation of protein homooligomerization
          positive regulation of mitochondrial fission
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27844462 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27844461 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P232470 AK171444.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P232470 BC088984.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P232470 NM_001009927.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P232470 AK156917.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P232470 AK171444.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P232470 BC088984.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P232470 NM_001009927.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P232470 AK156917.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 522076 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 628103 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 558475 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 571661 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P232470 AK171444.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P232470 BC088984.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P232470 NM_001009927.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P232470 AK156917.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P232470 AK171444.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P232470 BC088984.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P232470 NM_001009927.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P232470 AK156917.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27844467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27844475 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27844468 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27844489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27844478 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27844473 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27844464 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27844476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27844486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27844480 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27844469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27844470 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27844488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27844462 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27844491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27844483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27844465 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27844494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27844461 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27844481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27844466 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02339559 - -
          Whole Mouse Genome G4122A A_52_P232470 AK171444.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P232470 BC088984.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P232470 NM_001009927.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P232470 AK156917.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P232470 AK171444.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P232470 BC088984.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P232470 NM_001009927.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P232470 AK156917.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09040837 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09040836 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27844461 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27844462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27844464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27844501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09040837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27844497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27844470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27844457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27844491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27844481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27844476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08986718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09106109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09297689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09297687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27844486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09106108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27844464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27844475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09147500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27844498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09147489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27844473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27844488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27844467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27844496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27844508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27844459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09040846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09147498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27844465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27844506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27844466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09147496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27844478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09406294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27844468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09297688 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10376716 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376720 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376719 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376718 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376717 - -
          Affymetrix HT MG-430 PM Array Plate 1434072_PM_at - -
          Mouse Genome 430 2.0 Array 1434072_at - -
          Mouse Genome 430 2.0 Array 1434072_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 237781_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 237781_at - -
          Mouse Expression Set 430 1434072_PM_at - -
          Mouse Expression Set 430 1434072_at - -
          Murine Genome U74v2 Set 128879_f_at - -
          Murine Genome U74v2 Set 128879_PM_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1434072_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1434072_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 575656 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2534170 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2995370 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2995370 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline