Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01248558_g1
          See other TMEM256 GE Assays ›
          Gene Symbol:
          Tmem256
          Gene Name:
          transmembrane protein 256
          Gene Aliases:
          1810027O10Rik, 3110009M16Rik
          Chromosome Location:
          Chr.11: 69838525 - 69839558 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01248558_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem256
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026982.1 NP_081258.1 3 - 4 244 67
          GenBank mRNA AK007623.1 - 3 - 4 219 67
          AK014035.1 - 1 - 2 712 67
          BC048542.1 - 3 - 4 246 67
          BC117077.1 - 3 - 4 244 40126396 67
          BC117079.1 - 3 - 4 244 40126398 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000925.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P100932 BC117079.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P100932 AK014035.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P100932 BC048542.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P100932 BC117077.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P100932 NM_026982.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 453191 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P100932 BC117079.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P100932 AK014035.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P100932 BC048542.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P100932 BC117077.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P100932 NM_026982.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P651942 BC117079.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P651942 BC048542.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P651942 BC117077.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P651942 NM_026982.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P651942 AK007623.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27897445 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27897438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27897436 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27897448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27897429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08995102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08995106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27897450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27897433 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10377607 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377610 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377608 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377609 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377606 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377611 - -
          Affymetrix HT MG-430 PM Array Plate 1430985_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10377605 - -
          Mouse Genome 430 2.0 Array 1430985_at - -
          Mouse Genome 430 2.0 Array 1430985_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69186_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69186_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100007877.mm.1 NM_026982.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007877.mm.1 NM_026982.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007877.mm.1 TC1100000925.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100007878.mm.1 NM_026982.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007878.mm.1 NM_026982.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007878.mm.1 TC1100000925.mm.1 MTA TC
          Mouse Expression Set 430 1430985_PM_at - -
          Mouse Expression Set 430 1430985_at - -
          Murine Genome U74v2 Set 165983_PM_f_at - -
          Murine Genome U74v2 Set 165983_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1430985_at - -
          Affymetrix HT Mouse Genome 430B Array 1430985_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 907414 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 7000315 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2875089 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2875089 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline