Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01248720_m1
          See other CCDC103 GE Assays ›
          Gene Symbol:
          Ccdc103
          Gene Name:
          coiled-coil domain containing 103
          Gene Aliases:
          1700039D13Rik
          Chromosome Location:
          Chr.11: 102881244 - 102885215 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01248720_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccdc103
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028492.2 NP_082768.1 4 - 5 437 63
          GenBank mRNA AK006639.1 - 4 - 5 389 63
          AK032002.1 - 4 - 5 438 63
          BC119102.1 - 4 - 5 436 8733851 63
          BC119104.1 - 4 - 5 436 8733853 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001702.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccdc103

          Gene Ontology Categories:

          Function(s) Process(es)

          heart looping
          cilium movement
          cell projection organization
          outer dynein arm assembly
          inner dynein arm assembly
          axonemal dynein complex assembly
          determination of digestive tract left/right asymmetry
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2046288 NM_028492.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2046288 AK032002.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161463 BC144894.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161463 BC119104.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161463 NM_028492.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161463 AK006639.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161463 AK032002.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161463 BC144894.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161463 BC119104.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161463 NM_028492.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161463 AK006639.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161463 AK032002.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 191959 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 482666 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 269390 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 655942 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 851117 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 606958 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 831004 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 172264 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 159490 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 51359 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 144463 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 146716 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147343 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166443 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P176095 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161463 BC144894.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161463 BC119104.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161463 NM_028492.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161463 AK006639.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161463 AK032002.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161463 BC144894.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161463 BC119104.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161463 NM_028492.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161463 AK006639.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161463 AK032002.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2046288 NM_028492.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2046288 AK032002.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166443 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P147343 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P176095 - -
          Whole Mouse Genome G4122A A_51_P161463 BC144894.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P161463 BC119104.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P161463 NM_028492.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P161463 AK006639.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P161463 AK032002.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P161463 BC144894.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P161463 BC119104.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P161463 NM_028492.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P161463 AK006639.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P161463 AK032002.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P405200 BC144894.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P405200 BC119104.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P405200 NM_028492.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P405200 AK006639.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P405200 AK032002.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166443 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P147343 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P176095 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09067680 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09067685 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09067672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28081997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28081998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28082103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28082029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28082014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28082035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28082040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28082019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28082003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09720883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28082043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09589226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28082038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28082012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28082008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28082001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28082026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28082020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09572012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28082005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28082013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28082025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28082032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28082016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28082023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28082027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09572002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28082009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28082045 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10381656 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381660 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381659 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381657 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381655 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381658 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381654 - -
          Affymetrix HT MG-430 PM Array Plate 1453274_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10381653 - -
          Mouse Genome 430 2.0 Array 1453274_PM_at - -
          Mouse Genome 430 2.0 Array 1453274_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73293_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73293_at - -
          Affymetrix Mouse Clariom S Array PSR1100016039.mm.1 NM_028492.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016039.mm.1 NM_028492.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016039.mm.1 TC1100001702.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100016038.mm.1 NM_028492.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016038.mm.1 NM_028492.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016038.mm.1 TC1100001702.mm.1 MTA TC
          Mouse Expression Set 430 1453274_PM_at - -
          Mouse Expression Set 430 1453274_at - -
          Affymetrix HT Mouse Genome 430A Array 1421585_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421585_at - -
          Affymetrix HT Mouse Genome 430B Array 1453274_at - -
          Affymetrix HT Mouse Genome 430B Array 1453274_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 575715 - -
          ABI Mouse Genome Survey Microarray 593813 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1224090 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1224090 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline