Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01248965_m1
          See other CES2F GE Assays ›
          Gene Symbol:
          Ces2f
          Gene Name:
          carboxylesterase 2F
          Gene Aliases:
          2310038E17Rik, A430088E12
          Chromosome Location:
          Chr.8: 104947355 - 104961130 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01248965_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ces2f
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001079865.2 NP_001073334.2 12 - 13 1723 96
          GenBank mRNA BC117742.1 - 12 - 13 1723 40090393 96
          BC117743.1 - 10 - 11 1330 40090395 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001197.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          esterase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          carboxylic ester hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2101536 AK040349.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2101536 BC117742.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2101536 BC117743.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2101536 NM_001079865.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 256357 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 298826 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 350443 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 22154 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 515377 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 56682 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 14882 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2101536 AK040349.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2101536 BC117742.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2101536 BC117743.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2101536 NM_001079865.2 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26131039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130916 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06950906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07279908 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06700584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130908 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06960056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06700577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06806680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06700580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06950912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06780487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07063152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06700565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06806677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06780490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130913 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06950911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06950913 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07271020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06806676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07063148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130932 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10574561 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574566 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574571 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574568 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574563 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574565 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574569 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574567 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574564 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574570 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574562 - -
          Affymetrix HT MG-430 PM Array Plate 1432976_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1432975_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10574560 - -
          Mouse Genome 430 2.0 Array 1432975_at - -
          Mouse Genome 430 2.0 Array 1432976_at - -
          Mouse Genome 430 2.0 Array 1432975_PM_at - -
          Mouse Genome 430 2.0 Array 1432976_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800009350.mm.1 NM_001079865.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009350.mm.1 NM_001079865.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009350.mm.1 TC0800001197.mm.1 MTA TC
          Mouse Expression Set 430 1432975_PM_at - -
          Mouse Expression Set 430 1432976_PM_at - -
          Mouse Expression Set 430 1432976_at - -
          Mouse Expression Set 430 1432975_at - -
          Affymetrix HT Mouse Genome 430B Array 1432976_at - -
          Affymetrix HT Mouse Genome 430B Array 1432975_at - -
          Affymetrix HT Mouse Genome 430B Array 1432976_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432975_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline