Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01249295_g1
          See other MYO15B GE Assays ›
          Gene Symbol:
          Myo15b
          Gene Name:
          myosin XVB
          Gene Aliases:
          E330039G21Rik, Gm688, Gm886, mKIAA1783
          Chromosome Location:
          Chr.11: 115858391 - 115892594 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01249295_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Myo15b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK087906.2 - 16 - 17 100
          BC024506.1 - 11 - 12 1182 99
          BC051090.1 - 14 - 15 1450 99
          BF581819.1 - 3 - 4 360 99

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

          Panther Classification:

          Molecular Function -

          actin binding motor protein

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P371816 AK087906.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P371816 AK087906.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168264 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163765 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146500 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P117516 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996821 AK173262.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996821 AA474528.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P371816 AK087906.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P371816 AK087906.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2151695 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P168264 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P146500 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P117516 - -
          Whole Mouse Genome G4122A A_52_P632878 AK173262.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P632878 BB072116.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P168264 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P146500 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P117516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28155965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28155955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28155959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09005444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09171460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28155957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09171456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28155969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28155971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28155964 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10382682 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382680 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382685 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382679 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382688 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382690 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382686 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382689 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382687 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382681 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382678 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382683 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382691 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382684 - -
          Affymetrix HT MG-430 PM Array Plate 1435979_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1427203_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10382653 - -
          Mouse Genome 430 2.0 Array 1427203_at - -
          Mouse Genome 430 2.0 Array 1435979_PM_a_at - -
          Mouse Genome 430 2.0 Array 1435979_a_at - -
          Mouse Genome 430 2.0 Array 1427203_PM_at - -
          Mouse Expression Set 430 1427203_at - -
          Mouse Expression Set 430 1435979_a_at - -
          Mouse Expression Set 430 1427203_PM_at - -
          Mouse Expression Set 430 1435979_PM_a_at - -
          Murine Genome U74v2 Set 135494_PM_at - -
          Murine Genome U74v2 Set 135494_at - -
          Mouse Genome 430A 2.0 Array 1435979_a_at - -
          Mouse Genome 430A 2.0 Array 1435979_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1427203_at - -
          Mouse Genome 430A 2.0 Array 1427203_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1435979_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1427203_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427203_at - -
          Affymetrix HT Mouse Genome 430A Array 1435979_a_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1219746 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline