Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01249536_m1
          See other KCNG1 GE Assays ›
          Gene Symbol:
          Kcng1
          Gene Name:
          potassium voltage-gated channel, subfamily G, member 1
          Gene Aliases:
          AW536275, OTTMUSG00000016048
          Chromosome Location:
          Chr.2: 168260110 - 168283440 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01249536_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kcng1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081134.1 NP_001074603.1 1 - 2 861 87
          GenBank mRNA BC147251.1 - 1 - 2 793 9056302 87
          BC147252.1 - 1 - 2 793 9056297 87
          BC147715.1 - 1 - 2 793 9087474 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200005214.mm.1

          Set Membership:

          Amplicon length between 86 and 100 Ion Channel 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          potassium ion transport
          regulation of ion transmembrane transport
          protein homooligomerization
          transmembrane transport
          potassium ion transmembrane transport
          ion channel activity
          voltage-gated ion channel activity
          voltage-gated potassium channel activity
          delayed rectifier potassium channel activity
          potassium channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2073503 BC147715.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2073503 NM_001081134.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2073503 BC147251.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2073503 BC147252.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2055189 NM_001081134.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 94476 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 953346 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 389002 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 86948 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2073503 BC147715.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2073503 NM_001081134.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2073503 BC147251.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2073503 BC147252.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2055189 NM_001081134.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21840379 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01519315 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840463 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840453 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840420 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840400 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840468 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01519321 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840437 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840456 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01399941 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840434 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01519331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840427 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840426 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840406 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840424 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840447 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01261323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01569721 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840442 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840388 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840398 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840438 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01918657 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01850242 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01399942 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840421 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01569741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01918660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840458 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840465 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01261325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840472 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840436 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01569718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01263006 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01586878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01709946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01569727 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840412 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840394 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840419 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01399946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840408 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840413 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01519328 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01569729 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840475 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840445 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840428 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01569717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02117289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840443 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01850237 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840422 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840452 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01617047 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840451 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01891852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21840379 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01882289 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10489960 - -
          Affymetrix Mouse Gene 1.0 ST Array 10489959 - -
          Affymetrix HT MG-430 PM Array Plate 1459656_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1457052_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10489958 - -
          Mouse Genome 430 2.0 Array 1459656_PM_at - -
          Mouse Genome 430 2.0 Array 1459656_at - -
          Affymetrix Mouse Clariom S Array PSR0200044626.mm.1 NM_001081134.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044626.mm.1 NM_001081134.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044626.mm.1 TC0200005214.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200044627.mm.1 NM_001081134.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044627.mm.1 NM_001081134.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044627.mm.1 TC0200005214.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200044631.mm.1 NM_001081134.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044631.mm.1 NM_001081134.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044631.mm.1 TC0200005214.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200044628.mm.1 NM_001081134.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044628.mm.1 NM_001081134.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044628.mm.1 TC0200005214.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200044629.mm.1 NM_001081134.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044629.mm.1 NM_001081134.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044629.mm.1 TC0200005214.mm.1 MTA TC
          Mouse Expression Set 430 1459656_at - -
          Mouse Expression Set 430 1459656_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1459656_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1457052_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1459656_at - -
          Affymetrix HT Mouse Genome 430B Array 1457052_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 613521 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2990269 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2990269 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline