Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01250812_g1
          See other SLC16A11 GE Assays ›
          Gene Symbol:
          Slc16a11
          Gene Name:
          solute carrier family 16 (monocarboxylic acid transporters), member 11
          Gene Aliases:
          AB041591, MNCb-2717
          Chromosome Location:
          Chr.11: 70213910 - 70216414 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01250812_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc16a11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153081.3 NP_694721.2 2 - 3 563 143
          GenBank mRNA BC130255.1 - 1 - 2 66 143

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
          This assay is in the 5'UTR region of the BC130255.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc16a11

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          transport
          monocarboxylic acid transport
          ion transmembrane transport
          transmembrane transport
          anion transmembrane transport
          organic acid transmembrane transport
          molecular_function
          monocarboxylic acid transmembrane transporter activity
          symporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27899468 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27899471 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27899475 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27899474 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27899470 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27899473 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2061338 BE990575.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2061338 NM_153081.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2061338 AB041591.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 804246 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124940 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2061338 BE990575.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2061338 NM_153081.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2061338 AB041591.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899475 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899478 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899447 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899452 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899463 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899480 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899458 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899465 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899470 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899471 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899466 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899457 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899468 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899477 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899479 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899473 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899472 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899462 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899461 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899455 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P124940 - -
          Whole Mouse Genome G4122A A_51_P489720 BE990575.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P489720 NM_153081.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P489720 AB041591.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139847 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P124940 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09641177 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09701649 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09701639 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09701632 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09701629 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09641182 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09701637 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27899470 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27899473 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27899471 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27899468 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27899475 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27899474 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10377798 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377796 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377791 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377799 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377797 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377793 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377794 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377795 - -
          Affymetrix HT MG-430 PM Array Plate 1427107_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10377790 - -
          Mouse Genome 430 2.0 Array 1427107_PM_at - -
          Mouse Genome 430 2.0 Array 1427107_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 216867_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 216867_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100008291.mm.1 NM_153081.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008291.mm.1 NM_153081.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008291.mm.1 TC1100000943.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008290.mm.1 NM_153081.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008290.mm.1 NM_153081.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008290.mm.1 TC1100000943.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008286.mm.1 NM_153081.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008286.mm.1 NM_153081.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008286.mm.1 TC1100000943.mm.1 MTA TC
          Mouse Expression Set 430 1427107_at - -
          Mouse Expression Set 430 1427107_PM_at - -
          Murine Genome U74v2 Set 139487_at - -
          Murine Genome U74v2 Set 139487_PM_at - -
          Mouse Genome 430A 2.0 Array 1427107_at - -
          Mouse Genome 430A 2.0 Array 1427107_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427107_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427107_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 491270 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6380369 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006380369 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2721413 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2721413 - -
          mouse-6 v1.1 (Illumina) scl41341.3_8-S - -
          Illumina Sentrix mouse-6.v1 bead array 6380369 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl41341.3_8-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6380369 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2721413 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2721413 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline