Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01250908_m1
          See other BEAN1 GE Assays ›
          Gene Symbol:
          Bean1
          Gene Name:
          brain expressed, associated with Nedd4, 1
          Gene Aliases:
          Bean
          Chromosome Location:
          Chr.8: 104170376 - 104219098 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01250908_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Bean1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001141922.1 NP_001135394.1 6 - 7 1215 105
          NM_001141924.1 NP_001135396.1 4 - 5 998 105
          NM_001141925.1 NP_001135397.1 4 - 5 1000 105
          GenBank mRNA AF240460.1 - 2 - 3 395 105
          AK134633.1 - 4 - 5 1000 105
          BC030426.1 - 4 - 5 1078 105
          BC138184.1 - 6 - 7 1144 8861203 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001177.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P111962 AK134633.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P111962 NM_001141925.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P111962 AF240460.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P111962 AI851802.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P111962 NM_001141922.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P111962 AK134633.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P111962 NM_001141925.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P111962 AF240460.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P111962 AI851802.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P111962 NM_001141922.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 687570 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904204 AK011228.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904204 AK012467.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904204 AK013030.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904204 AK018391.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904204 AK164298.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904204 BC029246.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904204 BC037075.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904204 BC084681.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904204 NM_026529.4 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 274024 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 568179 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923136 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 749518 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 414768 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 415 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 680112 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 973306 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 51381 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 132890 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 607055 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 403464 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 300746 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 604757 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 729658 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 542986 BC022177.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 664062 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 309855 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 268018 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 432233 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 413791 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 296710 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 581123 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 841055 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 701141 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 564428 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 472881 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 750756 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 207267 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 16285 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 137132 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 48748 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 33969 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 53624 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P151091 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171981 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P111962 AK134633.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P111962 NM_001141925.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P111962 AF240460.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P111962 AI851802.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P111962 NM_001141922.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P111962 AK134633.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P111962 NM_001141925.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P111962 AF240460.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P111962 AI851802.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P111962 NM_001141922.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06623467 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P151091 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P171981 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01815950 - -
          Whole Mouse Genome G4122A A_51_P111962 AK134633.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P111962 NM_001141925.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P111962 AF240460.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P111962 AI851802.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P111962 NM_001141922.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P111962 AK134633.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P111962 NM_001141925.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P111962 AF240460.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P111962 AI851802.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P111962 NM_001141922.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P327110 AK134633.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P327110 BC030426.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P327110 NM_001141925.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P327110 AF240460.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P327110 NM_001141922.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P151091 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P171981 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01815961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06705302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07253335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127113 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07389672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06776911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06682046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07389671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06670538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07060217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07426780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07338653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07282438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07126858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07144995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07285078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07097786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06860241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07285083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06860240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07426777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07060219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06715597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07338651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07102652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07389666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06990011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07007377 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26127114 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10574455 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574453 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574454 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574452 - -
          Affymetrix HT MG-430 PM Array Plate 1452451_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10574451 - -
          Mouse Genome 430 2.0 Array 1452451_PM_at - -
          Mouse Genome 430 2.0 Array 1452451_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 65115_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 65115_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800009152.mm.1 NM_001141922.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009152.mm.1 NM_001141922.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009152.mm.1 NM_001141924.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009152.mm.1 NM_001141924.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009152.mm.1 NM_001141925.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009152.mm.1 NM_001141925.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009152.mm.1 TC0800001177.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009154.mm.1 NM_001141922.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009154.mm.1 NM_001141922.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009154.mm.1 NM_001141924.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009154.mm.1 NM_001141924.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009154.mm.1 NM_001141925.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009154.mm.1 NM_001141925.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009154.mm.1 TC0800001177.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009150.mm.1 NM_001141922.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009150.mm.1 NM_001141922.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009150.mm.1 NM_001141925.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009150.mm.1 NM_001141925.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009150.mm.1 TC0800001177.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009147.mm.1 NM_001141922.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009147.mm.1 NM_001141922.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009147.mm.1 TC0800001177.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009153.mm.1 NM_001141922.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009153.mm.1 NM_001141922.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009153.mm.1 NM_001141924.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009153.mm.1 NM_001141924.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009153.mm.1 NM_001141925.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009153.mm.1 NM_001141925.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009153.mm.1 TC0800001177.mm.1 MTA TC
          Mouse Expression Set 430 1452451_at - -
          Mouse Expression Set 430 1452451_PM_at - -
          Murine Genome U74v2 Set 134382_at - -
          Murine Genome U74v2 Set 137137_PM_at - -
          Murine Genome U74v2 Set 134382_PM_at - -
          Murine Genome U74v2 Set 137137_at - -
          Mouse Genome 430A 2.0 Array 1452451_at - -
          Mouse Genome 430A 2.0 Array 1452451_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452451_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452451_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 533486 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2698221 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0