Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01251252_m1
          See other NRSN2 GE Assays ›
          Gene Symbol:
          Nrsn2
          Gene Name:
          neurensin 2
          Gene Aliases:
          Gm123, Neurensin-2
          Chromosome Location:
          Chr.2: 152368755 - 152377153 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01251252_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nrsn2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009948.1 NP_001009948.1 2 - 3 213 64
          GenBank mRNA AK140190.1 - 2 - 3 230 64
          BC088982.1 - 2 - 3 245 5700110 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004897.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nrsn2

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2009427 BC088982.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2009427 NM_001009948.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 182873 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 413995 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 874900 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 672878 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 842651 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 140747 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158261 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2009427 BC088982.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2009427 NM_001009948.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749048 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749018 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158261 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158261 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02067195 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02067205 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01808411 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02067193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21748936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21748945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21748986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21748969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21748996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21748942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21748939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21748943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21748957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21748999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21748970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21748984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21748982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21748993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21748998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21748967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21748977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21748994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21748987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21748975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21748946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21748947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21748958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21748944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21748979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21748976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21748972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21748989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21748987 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749042 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21748998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749009 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21748941 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01659841 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01555369 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01404963 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21748993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749031 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749034 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21748989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01555372 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749032 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01430226 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749022 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749004 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749024 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01253077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01425054 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749050 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01555370 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21748994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749001 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749026 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01476788 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749036 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01582752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21748940 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01659843 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02067203 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21748945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21748984 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21748946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749029 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01207642 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21748982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01207639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01253082 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21748999 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749017 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02272512 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749007 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749037 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21748996 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01430225 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749044 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01929350 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10477104 - -
          Affymetrix Mouse Gene 1.0 ST Array 10488618 - -
          Affymetrix Mouse Gene 1.0 ST Array 10488619 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477106 - -
          Affymetrix Mouse Gene 1.0 ST Array 10488622 - -
          Affymetrix Mouse Gene 1.0 ST Array 10488621 - -
          Affymetrix Mouse Gene 1.0 ST Array 10488620 - -
          Affymetrix HT MG-430 PM Array Plate 1434640_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1447717_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10488617 - -
          Affymetrix Mouse Gene 1.1 ST Array 10477103 - -
          Mouse Genome 430 2.0 Array 1447717_x_at - -
          Mouse Genome 430 2.0 Array 1447717_PM_x_at - -
          Mouse Genome 430 2.0 Array 1434640_PM_at - -
          Mouse Genome 430 2.0 Array 1434640_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 228777_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 228777_at - -
          Affymetrix Mouse Clariom S Array PSR0200041603.mm.1 TC0200004897.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200041600.mm.1 NM_001009948.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041600.mm.1 NM_001009948.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041600.mm.1 TC0200004897.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200041601.mm.1 NM_001009948.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041601.mm.1 NM_001009948.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041601.mm.1 TC0200004897.mm.1 MTA TC
          Mouse Expression Set 430 1434640_at - -
          Mouse Expression Set 430 1434640_PM_at - -
          Mouse Expression Set 430 1447717_x_at - -
          Mouse Expression Set 430 1447717_PM_x_at - -
          Murine Genome U74v2 Set 130500_at - -
          Murine Genome U74v2 Set 130500_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1447717_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1447717_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1434640_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1434640_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 555023 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1231743 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2812599 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2812599 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0