Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01252036_m1
          See other TSEN54 GE Assays ›
          Gene Symbol:
          Tsen54
          Gene Name:
          tRNA splicing endonuclease 54 homolog (S. cerevisiae)
          Gene Aliases:
          0610034P02Rik
          Chromosome Location:
          Chr.11: 115813743 - 115823102 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01252036_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tsen54
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029557.1 NP_083833.1 8 - 9 1272 75
          GenBank mRNA AK002758.1 - 8 - 9 1257 75
          AK089005.1 - 8 - 9 1239 75
          BC051543.1 - 1 - 2 253 75
          BC117762.1 - 2 - 3 327 75
          BC126953.1 - 7 - 8 1396 75

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the BC117762.1 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          RNA metabolism protein endoribonuclease nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA-type intron splice site recognition and cleavage
          tRNA splicing, via endonucleolytic cleavage and ligation
          mRNA processing
          tRNA processing
          tRNA-intron endonuclease activity
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28155492 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28155497 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28155496 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28155493 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28155494 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28155495 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2198364 BC126953.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2198364 BC028453.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2198364 BC051543.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2198364 BC117762.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2198364 NM_029557.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963687 AK197075.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963687 BC028453.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963687 NM_029557.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963687 AK002758.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 789391 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 43294 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 775720 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 449046 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 26039 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 129079 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118519 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2198364 BC126953.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2198364 BC028453.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2198364 BC051543.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2198364 BC117762.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2198364 NM_029557.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963687 AK197075.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963687 BC028453.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963687 NM_029557.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963687 AK002758.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P434953 AK089005.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P434953 AK197075.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P434953 BC028453.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P434953 BC051543.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P434953 NM_029557.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P434953 AK089005.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P434953 AK197075.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P434953 BC028453.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P434953 BC051543.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P434953 NM_029557.1 mouse_transcriptome
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28155493 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28155497 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28155494 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28155495 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28155492 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28155496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28155419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28155520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28155546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28155526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09610352 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28155533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28155503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09610350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28155488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09610339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28155505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28155499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09333086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09587158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09327343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28155501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28155489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28155513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28155542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28155517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28155537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28155535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28155540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09245207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28155522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28155530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09702058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09448462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09610355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28155506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28155519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09610357 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10382613 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382624 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382616 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382620 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382611 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382615 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382619 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382617 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382623 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382622 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382618 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382621 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382614 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382612 - -
          Affymetrix HT MG-430 PM Array Plate 1429529_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10382610 - -
          Mouse Genome 430 2.0 Array 1429529_at - -
          Mouse Genome 430 2.0 Array 1429529_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76265_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76265_at - -
          Affymetrix Mouse Clariom S Array PSR1100017985.mm.1 NM_029557.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017985.mm.1 NM_029557.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017985.mm.1 TC1100001907.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100017986.mm.1 NM_029557.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017986.mm.1 NM_029557.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017986.mm.1 TC1100001907.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100017984.mm.1 NM_029557.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017984.mm.1 NM_029557.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017984.mm.1 TC1100001907.mm.1 MTA TC
          Mouse Expression Set 430 1429529_PM_at - -
          Mouse Expression Set 430 1429529_at - -
          Affymetrix HT Mouse Genome 430B Array 1429529_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429529_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 340289 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2729008 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2729008 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline