Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01252392_m1
          See other FAM89A GE Assays ›
          Gene Symbol:
          Fam89a
          Gene Name:
          family with sequence similarity 89, member A
          Gene Aliases:
          2310031A18Rik
          Chromosome Location:
          Chr.8: 124740257 - 124751809 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01252392_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam89a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081120.1 NP_001074589.1 1 - 2 263 66
          GenBank mRNA BC023460.1 - 1 - 2 104 66
          BC115734.1 - 1 - 2 122 40106966 66
          BC115819.1 - 1 - 2 122 40106968 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800003175.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26247506 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26247504 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26247505 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043627 NM_001081120.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043627 AK009550.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043627 BC023460.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2081116 BC115819.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2081116 BC115734.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 927138 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 714347 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 788761 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 458088 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 104002 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 192646 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 87869 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 885748 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 77483 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 137137 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142904 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168032 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043627 NM_001081120.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043627 AK009550.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043627 BC023460.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2081116 BC115819.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2081116 BC115734.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142904 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P168032 - -
          Whole Mouse Genome G4122A A_52_P297765 NM_001081120.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P297765 AK009550.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P297765 BC023460.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P168032 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142904 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26247506 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26247505 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26247504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26247532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06790737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07124837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06940894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07219901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07059258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06685134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06800384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07327678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07124836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247472 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06940898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07116723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247461 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07126367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07030785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07219899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07116724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247479 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26247523 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10582696 - -
          Affymetrix Mouse Gene 1.0 ST Array 10582698 - -
          Affymetrix Mouse Gene 1.0 ST Array 10582697 - -
          Affymetrix Mouse Gene 1.0 ST Array 10582695 - -
          Affymetrix HT MG-430 PM Array Plate 1427356_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10582694 - -
          Mouse Genome 430 2.0 Array 1427356_at - -
          Mouse Genome 430 2.0 Array 1427356_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69627_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69627_at - -
          Affymetrix Mouse Clariom S Array PSR0800024414.mm.1 NM_001081120.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024414.mm.1 NM_001081120.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024414.mm.1 TC0800003175.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800024412.mm.1 NM_001081120.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024412.mm.1 NM_001081120.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024412.mm.1 TC0800003175.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800024411.mm.1 NM_001081120.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024411.mm.1 NM_001081120.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024411.mm.1 TC0800003175.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800024413.mm.1 NM_001081120.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024413.mm.1 NM_001081120.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024413.mm.1 TC0800003175.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800024415.mm.1 NM_001081120.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024415.mm.1 NM_001081120.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024415.mm.1 TC0800003175.mm.1 MTA TC
          Mouse Expression Set 430 1427356_PM_at - -
          Mouse Expression Set 430 1427356_at - -
          Mouse Genome 430A 2.0 Array 1427356_at - -
          Mouse Genome 430A 2.0 Array 1427356_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427356_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427356_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 828514 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2754940 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2754940 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline