Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01254931_m1
          See other LSM14A GE Assays ›
          Gene Symbol:
          Lsm14a
          Gene Name:
          LSM14 homolog A (SCD6, S. cerevisiae)
          Gene Aliases:
          2700023B17Rik, AA407828, AU017544, Tral
          Chromosome Location:
          Chr.7: 34344637 - 34389637 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01254931_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lsm14a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025948.2 NP_080224.1 10 - 11 1489 126
          GenBank mRNA AK003611.1 - 8 - 9 1049 126
          AK012276.1 - 9 - 10 1252 126
          BC031521.1 - 10 - 11 1489 2655161 126

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002832.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lsm14a

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of translation
          multicellular organism development
          cytoplasmic mRNA processing body assembly
          RIG-I signaling pathway
          defense response to virus
          positive regulation of type I interferon-mediated signaling pathway
          double-stranded DNA binding
          double-stranded RNA binding
          single-stranded RNA binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994388 BC031521.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994388 AK012276.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994388 AK013143.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994388 NM_025948.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994388 AK003611.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 326481 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 740957 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 99638 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 866272 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 831692 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 43985 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 639127 AF186107.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 639127 AK036842.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 639127 BC125240.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 639127 BC125247.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 639127 BC125261.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 639127 NM_016928.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 121130 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 747952 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 501321 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 494636 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 496348 AK165438.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 496348 AK220293.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 496348 NM_001080769.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 729496 AK018999.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 659652 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 43755 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 454358 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 972930 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 748884 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 90452 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 672845 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 33789 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 775635 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 914597 AK052534.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 914597 AK173317.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 914597 BC020319.2 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 914597 BC037000.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 914597 BC049856.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 914597 BC076633.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 914597 NM_172628.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 854287 BC016598.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 854287 BC017534.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 854287 BC029324.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 854287 BC039946.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 854287 NM_133829.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 32183 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 357922 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 97489 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 682908 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 399386 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 807042 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 11868 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 137005 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 153478 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 175593 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994388 BC031521.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994388 AK012276.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994388 AK013143.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994388 NM_025948.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994388 AK003611.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P320860 BC031521.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P320860 AK012276.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P320860 NM_025948.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P320860 AK003611.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103827 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10562389 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562385 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562384 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562387 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562383 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562391 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562388 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562390 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562392 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562386 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562393 - -
          Affymetrix HT MG-430 PM Array Plate 1428438_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1428437_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1457738_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1428436_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10562382 - -
          Mouse Genome 430 2.0 Array 1428438_PM_s_at - -
          Mouse Genome 430 2.0 Array 1428436_at - -
          Mouse Genome 430 2.0 Array 1428436_PM_at - -
          Mouse Genome 430 2.0 Array 1428437_at - -
          Mouse Genome 430 2.0 Array 1457738_at - -
          Mouse Genome 430 2.0 Array 1428437_PM_at - -
          Mouse Genome 430 2.0 Array 1457738_PM_at - -
          Mouse Genome 430 2.0 Array 1428438_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67070_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67070_at - -
          Affymetrix Mouse Clariom S Array PSR0700025342.mm.1 NM_025948.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025342.mm.1 NM_025948.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025342.mm.1 TC0700002832.mm.1 MTA TC
          Mouse Expression Set 430 1457738_at - -
          Mouse Expression Set 430 1457738_PM_at - -
          Mouse Expression Set 430 1428438_PM_s_at - -
          Mouse Expression Set 430 1428437_at - -
          Mouse Expression Set 430 1428437_PM_at - -
          Mouse Expression Set 430 1428438_s_at - -
          Mouse Expression Set 430 1428436_PM_at - -
          Mouse Expression Set 430 1428436_at - -
          Murine Genome U74v2 Set 160281_at - -
          Murine Genome U74v2 Set 160281_PM_at - -
          Mouse Genome 430A 2.0 Array 1457738_PM_at - -
          Mouse Genome 430A 2.0 Array 1457738_at - -
          Affymetrix HT Mouse Genome 430A Array 1457738_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1457738_at - -
          Affymetrix HT Mouse Genome 430B Array 1428437_at - -
          Affymetrix HT Mouse Genome 430B Array 1428437_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428438_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1428438_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1428436_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428436_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 391406 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 610168 - -
          Illumina mouse-6 v1.0 expression beadchip 610168 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000610168 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2748914 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2748914 - -
          mouse-6 v1.1 (Illumina) scl31481.11_594-S - -
          Illumina Sentrix mouse-6.v1 bead array 610168 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31481.11_594-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 610168 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2748914 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2748914 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl31481.11_594-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-fcf5f9d6d-msf5g:80/100.66.78.203:80.
          git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.51.0-2026.05.21-1.0