Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01259579_m1
          See other 2010013B24RIK GE Assays ›
          Gene Symbol:
          2010013B24Rik
          Gene Name:
          RIKEN cDNA 2010013B24 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01259579_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2010013B24Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK008222.1 - 1 - 2 322 123

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2219174 AK008222.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173386 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2219174 AK008222.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P173386 - -
          Whole Mouse Genome G4122A A_52_P1122623 AK008222.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125261 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P173386 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05086700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05824085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05079607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05638722 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05028704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05079605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05638727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05314177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05027704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05131028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05074524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05074515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05345484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05079606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05089431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05260783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922212 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05074522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05086702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24922204 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 357336 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106520332 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline