Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01260884_gH
          See other OBOX6 GE Assays ›
          Gene Symbol:
          Obox6
          Gene Name:
          oocyte specific homeobox 6
          Gene Aliases:
          D17Ertd599e
          Chromosome Location:
          Chr.7: 15833250 - 15839679 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01260884_gH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Obox6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145710.2 NP_663756.2 2 - 3 632 63
          GenBank mRNA AA547582.1 - 1 - 2 51 63
          BC128024.1 - 1 - 2 571 40136487 63
          BY731792.1 - 2 - 3 631 63
          CN721150.1 - 1 - 2 304 63

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Obox6

          Panther Classification:

          Molecular Function -

          homeodomain transcription factor

          Gene Ontology Categories:

          Function(s)

          biological_process

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P124638 NM_145710.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P124638 BG069935.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 267362 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 479236 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 299580 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 35211 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P175990 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P124638 NM_145710.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P124638 BG069935.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P198453 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P175990 - -
          Whole Mouse Genome G4122A A_51_P198453 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124638 NM_145710.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124638 BG069935.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127763 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P175990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876092 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10560167 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560166 - -
          Affymetrix Mouse Gene 1.0 ST Array 10446348 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560165 - -
          Affymetrix Mouse Gene 1.0 ST Array 10446347 - -
          Affymetrix HT MG-430 PM Array Plate 1440257_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10446346 - -
          Affymetrix Mouse Gene 1.1 ST Array 10560164 - -
          Mouse Genome 430 2.0 Array 1440257_PM_at - -
          Mouse Genome 430 2.0 Array 1440257_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 252830_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 252830_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700021188.mm.1 TC0700002412.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021188.mm.1 NM_145710.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021188.mm.1 NM_145710.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021189.mm.1 TC0700002412.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021189.mm.1 NM_145710.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021189.mm.1 NM_145710.2 Refseq
          Mouse Expression Set 430 1440257_at - -
          Mouse Expression Set 430 1440257_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440257_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440257_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 633913 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6020092 - -
          Illumina mouse-6 v1.0 expression beadchip 6020092 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006020092 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1220159 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1220159 - -
          mouse-6 v1.1 (Illumina) scl0252830.1_112-S - -
          Illumina Sentrix mouse-6.v1 bead array 6020092 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0252830.1_112-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6020092 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1220159 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1220159 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline