Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01266106_m1
          See other DUSP5 GE Assays ›
          Gene Symbol:
          Dusp5
          Gene Name:
          dual specificity phosphatase 5
          Gene Aliases:
          Gm337
          Chromosome Location:
          Chr.19: 53529318 - 53541322 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01266106_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dusp5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001085390.1 NP_001078859.1 3 - 4 748 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000756.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Protein Phosphatase 3' Most Recommended Assay Mus musculus Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          inactivation of MAPK activity
          endoderm formation
          protein dephosphorylation
          dephosphorylation
          negative regulation of transcription, DNA-templated
          protein tyrosine/serine/threonine phosphatase activity
          MAP kinase tyrosine/serine/threonine phosphatase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063237 NM_001085390.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 666529 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 267163 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 366752 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 479016 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 49329 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 60495 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 68101 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2063237 NM_001085390.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32183148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183114 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14466181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14171573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183099 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183084 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14466195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183100 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183106 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14284175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14198435 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183011 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14287733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14334313 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14408715 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183215 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183143 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183097 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14198440 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183096 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183122 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14284136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14163636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14383327 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183103 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14483652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14198442 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183224 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14408719 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14466191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183151 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183086 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14169658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183098 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14466180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14284171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14284135 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14383321 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183109 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183116 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14238911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183038 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183153 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14284168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14290139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183052 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183164 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183130 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183218 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14204119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14466192 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14334315 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14466175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32183217 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10463950 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463946 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463947 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463949 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463948 - -
          Affymetrix HT MG-430 PM Array Plate 1437199_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10463945 - -
          Mouse Genome 430 2.0 Array 1437199_PM_at - -
          Mouse Genome 430 2.0 Array 1437199_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 240672_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 240672_at - -
          Affymetrix Mouse Clariom S Array PSR1900006663.mm.1 NM_001085390.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900006663.mm.1 NM_001085390.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900006663.mm.1 TC1900000756.mm.1 MTA TC
          Mouse Expression Set 430 1437199_at - -
          Mouse Expression Set 430 1437199_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1255976 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0