Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01267202_m1
          See other CEP76 GE Assays ›
          Gene Symbol:
          Cep76
          Gene Name:
          centrosomal protein 76
          Gene Aliases:
          6230425F05Rik, 9630013E05
          Chromosome Location:
          Chr.18: 67617397 - 67641336 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01267202_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cep76
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081073.1 NP_001074542.1 6 - 7 966 65
          GenBank mRNA AK035879.1 - 5 - 6 746 65
          BC119216.1 - 6 - 7 847 8733965 65
          BC119218.1 - 6 - 7 847 8733967 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800001510.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of centriole replication
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P345316 AK035879.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P345316 AK165893.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P345316 NM_001081073.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P345316 AK035879.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P345316 AK165893.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P345316 NM_001081073.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 669959 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 388483 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 103542 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 187136 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897704 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 688185 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 81483 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 337297 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 795995 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 666189 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 242980 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 191952 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 719720 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 956705 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 233213 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 8553 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 44040 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 92852 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157275 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P345316 AK035879.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P345316 AK165893.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P345316 NM_001081073.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P345316 AK035879.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P345316 AK165893.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P345316 NM_001081073.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09037036 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157275 - -
          Whole Mouse Genome G4122A A_51_P345316 AK035879.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P345316 AK165893.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P345316 NM_001081073.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P345316 AK035879.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P345316 AK165893.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P345316 NM_001081073.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P387717 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157275 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14016802 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14016808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792813 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13947233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792819 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792957 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792930 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13614509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792798 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13662289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13807776 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14055835 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13818918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792932 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792788 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792823 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13740569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13985099 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792822 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792960 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13875870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13850366 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792835 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792973 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13737753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13740568 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13978748 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792935 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31792931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13947231 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10459588 - -
          Affymetrix Mouse Gene 1.0 ST Array 10459578 - -
          Affymetrix Mouse Gene 1.0 ST Array 10459589 - -
          Affymetrix Mouse Gene 1.0 ST Array 10459586 - -
          Affymetrix Mouse Gene 1.0 ST Array 10459580 - -
          Affymetrix Mouse Gene 1.0 ST Array 10459582 - -
          Affymetrix Mouse Gene 1.0 ST Array 10459587 - -
          Affymetrix Mouse Gene 1.0 ST Array 10459581 - -
          Affymetrix Mouse Gene 1.0 ST Array 10459583 - -
          Affymetrix Mouse Gene 1.0 ST Array 10459585 - -
          Affymetrix Mouse Gene 1.0 ST Array 10459577 - -
          Affymetrix Mouse Gene 1.0 ST Array 10459584 - -
          Affymetrix Mouse Gene 1.0 ST Array 10459579 - -
          Affymetrix HT MG-430 PM Array Plate 1437209_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1437254_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10459576 - -
          Mouse Genome 430 2.0 Array 1437254_at - -
          Mouse Genome 430 2.0 Array 1437254_PM_at - -
          Mouse Genome 430 2.0 Array 1437209_PM_at - -
          Mouse Genome 430 2.0 Array 1437209_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 225659_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 225659_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1800011089.mm.1 NM_001081073.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011089.mm.1 NM_001081073.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011089.mm.1 TC1800001510.mm.1 MTA TC
          Mouse Expression Set 430 1437209_PM_at - -
          Mouse Expression Set 430 1437209_at - -
          Mouse Expression Set 430 1437254_PM_at - -
          Mouse Expression Set 430 1437254_at - -
          Affymetrix HT Mouse Genome 430B Array 1437209_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1437209_at - -
          Affymetrix HT Mouse Genome 430B Array 1437254_at - -
          Affymetrix HT Mouse Genome 430B Array 1437254_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 656030 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1253298 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1253298 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline