Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01268045_s1
          See other FAM187A GE Assays ›
          Gene Symbol:
          Fam187a
          Gene Name:
          family with sequence similarity 187, member A
          Gene Aliases:
          4933439F11Rik
          Chromosome Location:
          Chr.11: 102885169 - 102886731 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01268045_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam187a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025766.2 NP_080042.1 1 1217 62
          GenBank mRNA AK016939.1 - 1 1218 62
          AK017119.1 - 1 1218 62
          BC116769.1 - 1 1063 40126088 62
          BC116771.1 - 1 1063 40126090 62

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fam187a

          Panther Classification:

          Molecular Function -

          immunoglobulin superfamily cell adhesion molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28082000 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2046293 AK017119.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2046293 NM_025766.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 800817 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161180 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2046293 AK017119.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2046293 NM_025766.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28082039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28082012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28082035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28082037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28082024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28082025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28082033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28082020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28082013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28082001 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28082032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28082014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28082030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28082016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28082038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28082003 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28082027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28082005 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28082017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28082011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28082021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28082019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28082009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28082031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28082022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28082029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28082040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28082008 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161180 - -
          Whole Mouse Genome G4122A A_51_P399517 AK016939.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P399517 AK017119.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P399517 BC116769.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P399517 BC116771.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P399517 NM_025766.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124026 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106059 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161180 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28082000 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10381662 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381663 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381661 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381660 - -
          Affymetrix Mouse Gene 1.1 ST Array 10381653 - -
          Mouse Genome 430 2.0 Array 1421585_at - -
          Mouse Genome 430 2.0 Array 1421585_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100016044.mm.1 NM_025766.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016044.mm.1 NM_025766.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016044.mm.1 TC1100001702.mm.1 MTA TC
          Mouse Expression Set 430 1421585_at - -
          Mouse Expression Set 430 1421585_PM_at - -
          Mouse Genome 430A 2.0 Array 1421585_at - -
          Mouse Genome 430A 2.0 Array 1421585_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 405208 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6650095 - -
          Illumina mouse-6 v1.0 expression beadchip 6650095 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006650095 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1244553 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1244553 - -
          mouse-6 v1.1 (Illumina) scl066784.1_299-S - -
          Illumina Sentrix mouse-6.v1 bead array 6650095 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl066784.1_299-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6650095 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1244553 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0