Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01271022_g1
          See other LSM5 GE Assays ›
          Gene Symbol:
          Lsm5
          Gene Name:
          LSM5 homolog, U6 small nuclear RNA associated (S. cerevisiae)
          Gene Aliases:
          2010208O10Rik, 2310034K10Rik
          Chromosome Location:
          Chr.6: 56701057 - 56704699 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01271022_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lsm5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025520.3 NP_079796.1 4 - 5 280 135
          GenBank mRNA AK008480.1 - 3 - 4 538 135
          AK009617.1 - 4 - 5 274 135
          AK011218.1 - 4 - 5 266 135
          BC048459.1 - 4 - 5 255 6601124 135
          BC061085.1 - 264 30295556 135
          BC132626.1 - 4 - 5 280 40130903 135
          BC132628.1 - 4 - 5 280 40130905 135
          CB204385.1 - 4 - 5 273 135

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lsm5

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA processing
          mRNA catabolic process
          RNA splicing
          RNA binding
          protein binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24475724 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24476403 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24476402 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24476400 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24476145 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24475727 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24475723 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24476404 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24476146 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24476143 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24476144 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24476401 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1967633 NM_025520.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1967633 AK008480.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1967633 AK009617.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1967633 AK011218.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1967633 BC061085.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2061437 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 360316 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 595577 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 81486 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119046 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1967633 NM_025520.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1967633 AK008480.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1967633 AK009617.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1967633 AK011218.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1967633 BC061085.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2061437 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P336790 AK009617.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24476387 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24476411 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24476409 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24476419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24476417 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24476394 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24476389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24476416 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24476392 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24476408 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24476415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24476421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24476386 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24476423 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24476412 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24476388 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24476396 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24476407 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24476422 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24476398 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24476401 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24476404 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24476395 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24476405 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24476391 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24476406 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P119046 - -
          Whole Mouse Genome G4122A A_51_P336790 AK009617.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112922 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P119046 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05378732 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05534706 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05378730 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05534700 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24476145 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24476403 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24476400 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24475727 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24476144 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24476402 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24476146 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24475724 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24476404 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24476143 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24476401 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24475723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24476426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476416 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05534700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476442 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476358 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476377 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476363 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05701550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05378729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476418 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476376 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476411 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05054873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05232707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476373 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05534708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05534698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476403 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476380 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476386 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476436 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05701542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05405874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05534710 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10353675 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544975 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544972 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544974 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445009 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544973 - -
          Affymetrix HT MG-430 PM Array Plate 1418656_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10544971 - -
          Affymetrix Mouse Gene 1.1 ST Array 10445008 - -
          Affymetrix Mouse Gene 1.1 ST Array 10353674 - -
          Mouse Genome 430 2.0 Array 1418656_at - -
          Mouse Genome 430 2.0 Array 1418656_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600018253.mm.1 TC0600002337.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600018253.mm.1 NM_025520.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018253.mm.1 NM_025520.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018256.mm.1 TC0600002337.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600018256.mm.1 NM_025520.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018256.mm.1 NM_025520.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018252.mm.1 TC0600002337.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600018252.mm.1 NM_025520.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018252.mm.1 NM_025520.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018255.mm.1 TC0600002337.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600018255.mm.1 NM_025520.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018255.mm.1 NM_025520.3 Refseq
          Mouse Expression Set 430 1418656_at - -
          Mouse Expression Set 430 1418656_PM_at - -
          Murine Genome U74v2 Set 94389_PM_at - -
          Murine Genome U74v2 Set 94389_at - -
          Mouse Genome 430A 2.0 Array 1418656_PM_at - -
          Mouse Genome 430A 2.0 Array 1418656_at - -
          Affymetrix HT Mouse Genome 430A Array 1418656_at - -
          Affymetrix HT Mouse Genome 430A Array 1418656_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 620133 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4540398 - -
          Illumina mouse-6 v1.0 expression beadchip 4540398 - -
          Illumina mouse-6 v1.0 expression beadchip 106860162 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004540398 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1257085 - -
          mouse-6 v1.1 (Illumina) scl0066373.2_111-S - -
          mouse-6 v1.1 (Illumina) scl28966.2_89-S - -
          Illumina Sentrix mouse-6.v1 bead array 4540398 - -
          Illumina Sentrix mouse-6.v1 bead array 106860162 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0066373.2_111-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106860162 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4540398 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2971946 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2624523 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1257085 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2971946 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline