Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01272722_m1
          See other SLC16A14 GE Assays ›
          Gene Symbol:
          Slc16a14
          Gene Name:
          solute carrier family 16 (monocarboxylic acid transporters), member 14
          Gene Aliases:
          1110004H10Rik
          Chromosome Location:
          Chr.1: 84905457 - 84935281 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01272722_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc16a14
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027921.1 NP_082197.1 4 - 5 1630 133
          GenBank mRNA AK003423.1 - 1 - 2 874 133
          AK049846.1 - 4 - 5 1682 133
          BC023456.1 - 4 - 5 1630 5041164 133

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100002702.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc16a14

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          monocarboxylic acid transport
          ion transmembrane transport
          transmembrane transport
          anion transmembrane transport
          organic acid transmembrane transport
          molecular_function
          monocarboxylic acid transmembrane transporter activity
          symporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1970144 AK049846.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P566129 BC023456.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P566129 AK049846.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P566129 NM_027921.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P566129 AK003423.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P566129 BC023456.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P566129 AK049846.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P566129 NM_027921.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P566129 AK003423.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 214312 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 528255 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 227873 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 859453 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 224088 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 775353 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 226024 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 469444 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 690802 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 270546 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 102670 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 256643 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 539605 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 907008 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 896736 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 48102 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 4883 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 78367 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 242 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142489 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123820 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P566129 BC023456.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P566129 AK049846.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P566129 NM_027921.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P566129 AK003423.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P566129 BC023456.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P566129 AK049846.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P566129 NM_027921.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P566129 AK003423.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03801712 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09499519 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142489 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09499518 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09499514 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03801715 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09499512 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09499523 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P123820 - -
          Whole Mouse Genome G4122A A_51_P204329 AK049846.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P566129 BC023456.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P566129 AK049846.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P566129 NM_027921.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P566129 AK003423.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P566129 BC023456.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P566129 AK049846.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P566129 NM_027921.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P566129 AK003423.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113905 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P123820 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142489 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32832579 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32832583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32832581 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32832584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32832577 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10356243 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356244 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356246 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356245 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356242 - -
          Affymetrix HT MG-430 PM Array Plate 1439661_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1430629_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10356240 - -
          Mouse Genome 430 2.0 Array 1430629_PM_at - -
          Mouse Genome 430 2.0 Array 1439661_PM_at - -
          Mouse Genome 430 2.0 Array 1439661_at - -
          Mouse Genome 430 2.0 Array 1430629_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71781_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71781_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100021920.mm.1 NM_027921.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100021920.mm.1 NM_027921.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100021920.mm.1 TC0100002702.mm.1 MTA TC
          Mouse Expression Set 430 1430629_PM_at - -
          Mouse Expression Set 430 1439661_at - -
          Mouse Expression Set 430 1430629_at - -
          Mouse Expression Set 430 1439661_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430629_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1439661_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1439661_at - -
          Affymetrix HT Mouse Genome 430B Array 1430629_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 810154 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106020347 - -
          Illumina mouse-6 v1.0 expression beadchip 106020347 - -
          Illumina mouse-6 v1.0 expression beadchip 3940270 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003940270 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1217704 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1217704 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1235245 - -
          mouse-6 v1.1 (Illumina) scl16531.6_458-S - -
          mouse-6 v1.1 (Illumina) scl0071781.2_8-S - -
          Illumina Sentrix mouse-6.v1 bead array 3940270 - -
          Illumina Sentrix mouse-6.v1 bead array 106020347 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0071781.2_8-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3940270 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106020347 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1217704 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1217704 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline