Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01272975_sH
          See other OLFR704 GE Assays ›
          Gene Symbol:
          Olfr704
          Gene Name:
          olfactory receptor 704
          Gene Aliases:
          MOR283-12P
          Chromosome Location:
          Chr.7: 106864982 - 106865929 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01272975_sH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr704
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011749.1 NP_001011749.1 1 637 103

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr704

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2080756 NM_001011749.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2080756 NM_001011749.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P231128 NM_001011749.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P231125 NM_001011749.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06593249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06315568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05916347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06045603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06493401 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396904 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566590 - -
          Affymetrix Mouse Gene 1.0 ST Array 10556032 - -
          Affymetrix Mouse Gene 1.1 ST Array 10556031 - -
          Affymetrix Mouse Gene 1.1 ST Array 10566589 - -
          Affymetrix Mouse Clariom S Array PSR0700013157.mm.1 TC0700001566.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700013157.mm.1 NM_001011749.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700013157.mm.1 NM_001011749.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 781797 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2926690 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2926690 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline