Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01274155_m1
          See other CHRNA10 GE Assays ›
          Gene Symbol:
          Chrna10
          Gene Name:
          cholinergic receptor, nicotinic, alpha polypeptide 10
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 102111262 - 102116828 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01274155_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Chrna10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081424.1 NP_001074893.1 1 - 2 456 70
          GenBank mRNA AK033068.1 - 1 - 2 457 70
          BC147245.1 - 1 - 2 171 9056293 70
          BC147246.1 - 1 - 2 171 9056310 70
          BC151033.1 - 1 - 2 171 9087946 70
          BC151035.1 - 1 - 2 171 9087948 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003853.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          ligand-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cytosolic calcium ion concentration
          synaptic transmission, cholinergic
          inner ear morphogenesis
          detection of mechanical stimulus involved in sensory perception of sound
          cation transmembrane transport
          acetylcholine-activated cation-selective channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1991244 NM_001081424.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1991244 AK033068.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1991244 AK009284.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 403413 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 382416 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 235572 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 67380 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1991244 NM_001081424.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1991244 AK033068.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1991244 AK009284.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P478774 NM_001081424.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P478774 AK033068.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P478774 AK009284.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123590 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121366 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555669 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555670 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566096 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566095 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566092 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566094 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566093 - -
          Affymetrix Mouse Gene 1.1 ST Array 10566091 - -
          Affymetrix Mouse Gene 1.1 ST Array 10555662 - -
          Affymetrix Mouse Clariom S Array PSR0700032567.mm.1 NM_001081424.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032567.mm.1 NM_001081424.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032567.mm.1 TC0700003853.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032568.mm.1 NM_001081424.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032568.mm.1 NM_001081424.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032568.mm.1 TC0700003853.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032569.mm.1 NM_001081424.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032569.mm.1 NM_001081424.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032569.mm.1 TC0700003853.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 481403 - -
          ABI Mouse Genome Survey Microarray 708116 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 102680053 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1252409 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222404 - -
          Illumina Sentrix mouse-6.v1 bead array 102680053 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102680053 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3073524 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3073524 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3073529 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline