Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01276228_m1
          See other PDILT GE Assays ›
          Gene Symbol:
          Pdilt
          Gene Name:
          protein disulfide isomerase-like, testis expressed
          Gene Aliases:
          1700007B13Rik
          Chromosome Location:
          Chr.7: 119486586 - 119528621 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01276228_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pdilt
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027943.1 NP_082219.1 11 - 12 1733 75
          GenBank mRNA AK005692.1 - 11 - 12 1733 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004184.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          protein folding
          multicellular organism development
          spermatogenesis
          spermatid development
          cell migration
          cell differentiation
          cell redox homeostasis
          protein disulfide isomerase activity
          protein binding
          isomerase activity
          peptidyl-proline 4-dioxygenase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P641282 AK005692.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P641282 NM_027943.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P641282 AK005692.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P641282 NM_027943.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 613175 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 440579 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 257313 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 963520 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 343057 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 159881 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 843446 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 72529 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 8775 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 880442 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 855934 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 949222 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 533482 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 1083 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 601182 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 627847 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 868535 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 305639 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 380059 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 831127 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 795004 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 775718 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 27059 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 32596 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123767 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107558 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P641282 AK005692.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P641282 NM_027943.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P641282 AK005692.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P641282 NM_027943.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107558 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P123767 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01618130 - -
          Whole Mouse Genome G4122A A_52_P641282 AK005692.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P641282 NM_027943.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P641282 AK005692.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P641282 NM_027943.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P123767 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05886444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05886438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25464067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25464073 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1454033_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1431867_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10567394 - -
          Mouse Genome 430 2.0 Array 1431867_a_at - -
          Mouse Genome 430 2.0 Array 1431867_PM_a_at - -
          Mouse Genome 430 2.0 Array 1454033_PM_at - -
          Mouse Genome 430 2.0 Array 1454033_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71830_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71830_at - -
          Affymetrix Mouse Clariom S Array PSR0700034806.mm.1 NM_027943.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700034806.mm.1 NM_027943.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700034806.mm.1 TC0700004184.mm.1 MTA TC
          Mouse Expression Set 430 1454033_PM_at - -
          Mouse Expression Set 430 1454033_at - -
          Mouse Expression Set 430 1431867_PM_a_at - -
          Mouse Expression Set 430 1431867_a_at - -
          Murine Genome U74v2 Set 168775_PM_at - -
          Murine Genome U74v2 Set 168775_at - -
          Affymetrix HT Mouse Genome 430B Array 1454033_at - -
          Affymetrix HT Mouse Genome 430B Array 1431867_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1454033_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431867_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 466511 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1236696 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1236696 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0