Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01279341_m1
          See other SAYSD1 GE Assays ›
          Gene Symbol:
          Saysd1
          Gene Name:
          SAYSVFN motif domain containing 1
          Gene Aliases:
          1810063B07Rik, 4930488P03Rik
          Chromosome Location:
          Chr.14: 20075636 - 20083172 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01279341_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Saysd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026209.1 NP_080485.1 1 - 2 298 104
          GenBank mRNA AK007941.1 - 253 104
          AK031748.1 - 1 - 2 224 104
          BC027644.1 - 1 - 2 219 1248085 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400001527.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29445010 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29445006 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29445011 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29445008 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29445005 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29445004 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29445007 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29445009 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2186180 AK031748.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2186180 CJ320425.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2186180 NM_026209.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 862047 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 276930 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 693398 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 166703 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731737 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146208 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P150446 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2186180 AK031748.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2186180 CJ320425.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2186180 NM_026209.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P595805 AK031748.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P595805 BC027644.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P595805 NM_026209.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P595805 AK007941.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P595805 AK021296.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109549 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29445006 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29445009 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29445008 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29445004 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29445005 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29445010 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29445007 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29445011 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29444965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29444961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29444949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29444944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29444966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11636279 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29444946 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11786718 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11786721 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29444952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11786712 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29444971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29444942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29444964 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29444968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11546779 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29444950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29444975 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10417795 - -
          Affymetrix Mouse Gene 1.0 ST Array 10417796 - -
          Affymetrix Mouse Gene 1.0 ST Array 10417797 - -
          Affymetrix HT MG-430 PM Array Plate 1442200_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1427905_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1430748_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10417794 - -
          Mouse Genome 430 2.0 Array 1427905_PM_at - -
          Mouse Genome 430 2.0 Array 1430748_at - -
          Mouse Genome 430 2.0 Array 1442200_at - -
          Mouse Genome 430 2.0 Array 1430748_PM_at - -
          Mouse Genome 430 2.0 Array 1442200_PM_at - -
          Mouse Genome 430 2.0 Array 1427905_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67509_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67509_at - -
          Affymetrix Mouse Clariom S Array PSR1400011361.mm.1 NM_026209.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400011361.mm.1 NM_026209.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400011361.mm.1 TC1400001527.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400011368.mm.1 NM_026209.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400011368.mm.1 NM_026209.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400011368.mm.1 TC1400001527.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400011366.mm.1 NM_026209.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400011366.mm.1 NM_026209.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400011366.mm.1 TC1400001527.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400011362.mm.1 NM_026209.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400011362.mm.1 NM_026209.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400011362.mm.1 TC1400001527.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400011367.mm.1 NM_026209.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400011367.mm.1 NM_026209.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400011367.mm.1 TC1400001527.mm.1 MTA TC
          Mouse Expression Set 430 1427905_PM_at - -
          Mouse Expression Set 430 1427905_at - -
          Mouse Expression Set 430 1442200_at - -
          Mouse Expression Set 430 1442200_PM_at - -
          Mouse Expression Set 430 1430748_at - -
          Mouse Expression Set 430 1430748_PM_at - -
          Murine Genome U74v2 Set 95011_at - -
          Murine Genome U74v2 Set 95011_PM_at - -
          Mouse Genome 430A 2.0 Array 1427905_at - -
          Mouse Genome 430A 2.0 Array 1427905_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427905_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427905_at - -
          Affymetrix HT Mouse Genome 430B Array 1442200_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430748_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430748_at - -
          Affymetrix HT Mouse Genome 430B Array 1442200_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 494115 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106770148 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1255493 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0