Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01284274_m1
          See other FAM179B GE Assays ›
          Gene Symbol:
          Fam179b
          Gene Name:
          family with sequence similarity 179, member B
          Gene Aliases:
          A430041B07Rik, BC062107, C130034K06, mKIAA0423
          Chromosome Location:
          Chr.12: 64965742 - 65022573 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01284274_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam179b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177805.3 NP_808473.2 15 - 16 4683 142
          GenBank mRNA AK148829.1 - 3 - 4 333 142
          AK172948.1 - 15 - 16 4611 142
          BC062107.1 - 5 - 6 715 142
          BC157958.1 - 15 - 16 4485 9088131 142
          BC158008.1 - 16 - 17 4635 9007316 142
          BC172117.1 - 15 - 16 4485 9054403 142

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200000523.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fam179b

          Panther Classification:

          Molecular Function -

          non-motor microtubule binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2067086 BC062107.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2067086 AK048104.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2067086 NM_177805.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2067091 BC157958.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2067091 BC158008.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2067091 BC172117.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2067091 AK172948.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2067091 AK141238.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 223248 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487358 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 475447 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 464129 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 646727 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 506667 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 430553 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 641686 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 80342 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 900170 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 517050 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 938954 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 837648 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 512450 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 66323 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 913331 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 842132 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 545940 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 288010 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 908934 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 956751 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 930124 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 450300 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 893493 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 674526 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 424172 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 882529 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 954554 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 959910 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 215714 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 78888 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 321120 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 143087 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 36285 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 130352 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 52327 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 4969 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127245 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159018 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P138573 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2067086 BC062107.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2067086 AK048104.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2067086 NM_177805.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2067091 BC157958.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2067091 BC158008.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2067091 BC172117.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2067091 AK172948.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2067091 AK141238.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P159018 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07645262 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P127245 - -
          Whole Mouse Genome G4122A A_51_P281447 BC157958.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281447 BC158008.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281447 BC172117.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281447 AK172948.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281447 AK141238.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P177988 BC157958.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P177988 BC158008.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P177988 BC172117.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P177988 AK172948.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P177988 AK084470.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105745 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P127245 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07645262 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P159018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493312 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493398 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493447 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493465 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10248486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493478 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493449 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10341096 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09936560 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493405 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493309 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493296 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493287 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493305 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493440 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09901087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493442 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493473 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493441 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493297 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493400 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493286 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10227680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493445 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493439 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493308 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493446 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10237740 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10169387 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493472 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493290 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493435 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493433 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493448 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493462 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493443 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493283 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493436 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493321 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493444 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493324 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493310 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493450 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10060477 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10225167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493318 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493476 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493480 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493455 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493292 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493307 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493315 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493293 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493302 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493467 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10153312 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493278 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493319 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493313 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493322 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493303 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493401 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493300 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493314 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493294 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493458 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493281 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28493452 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10396002 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395998 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395994 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395985 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395990 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395987 - -
          Affymetrix Mouse Gene 1.0 ST Array 10396001 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395997 - -
          Affymetrix Mouse Gene 1.0 ST Array 10396007 - -
          Affymetrix Mouse Gene 1.0 ST Array 10396003 - -
          Affymetrix Mouse Gene 1.0 ST Array 10396006 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395991 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395995 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395993 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395999 - -
          Affymetrix Mouse Gene 1.0 ST Array 10396005 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395989 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395988 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395996 - -
          Affymetrix Mouse Gene 1.0 ST Array 10396004 - -
          Affymetrix Mouse Gene 1.0 ST Array 10396000 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395992 - -
          Affymetrix HT MG-430 PM Array Plate 1458067_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1441095_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1434843_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10395984 - -
          Mouse Genome 430 2.0 Array 1441095_PM_at - -
          Mouse Genome 430 2.0 Array 1434843_at - -
          Mouse Genome 430 2.0 Array 1458067_PM_at - -
          Mouse Genome 430 2.0 Array 1441095_at - -
          Mouse Genome 430 2.0 Array 1434843_PM_at - -
          Mouse Genome 430 2.0 Array 1458067_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 328108_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 328108_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1200003623.mm.1 NM_177805.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003623.mm.1 NM_177805.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003623.mm.1 TC1200000523.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200003622.mm.1 NM_177805.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003622.mm.1 NM_177805.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003622.mm.1 TC1200000523.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200003621.mm.1 NM_177805.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003621.mm.1 NM_177805.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003621.mm.1 TC1200000523.mm.1 MTA TC
          Mouse Expression Set 430 1441095_PM_at - -
          Mouse Expression Set 430 1434843_at - -
          Mouse Expression Set 430 1458067_at - -
          Mouse Expression Set 430 1441095_at - -
          Mouse Expression Set 430 1434843_PM_at - -
          Mouse Expression Set 430 1458067_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1458067_at - -
          Affymetrix HT Mouse Genome 430B Array 1434843_at - -
          Affymetrix HT Mouse Genome 430B Array 1434843_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1441095_at - -
          Affymetrix HT Mouse Genome 430B Array 1458067_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1441095_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 337003 - -
          ABI Mouse Genome Survey Microarray 594195 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101170088 - -
          Illumina mouse-6 v1.0 expression beadchip 101170088 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1259781 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1251488 - -
          Illumina Sentrix mouse-6.v1 bead array 101170088 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101170088 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1259781 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline