Search
Search

Vcpkmt| Gene Symbol: Vcpkmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033236.2 | NP_001028408.2 | 1 - 2 | 306 | 115 | |
| GenBank mRNA | AK144832.1 | - | 1 - 2 | 269 | 115 | |
| BC138942.1 | - | 1 - 2 | 388 | 8861964 | 115 | |
| BC138943.1 | - | 1 - 2 | 388 | 8861965 | 115 | |
| BC145471.1 | - | 1 - 2 | 388 | 115 | ||
| BY095246.1 | - | 1 - 2 | 307 | 115 | ||
Set Membership: |
|