Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01294434_s1
          See other TEDDM1B GE Assays ›
          Gene Symbol:
          Teddm1b
          Gene Name:
          transmembrane epididymal protein 1B
          Gene Aliases:
          EG433365, Gm5531
          Chromosome Location:
          Chr.1: 153874345 - 153876871 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01294434_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Teddm1b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008426.3 NP_001008426.1 1 2484 86
          GenBank mRNA AK033834.1 - 1 2484 86

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Teddm1b

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P135003 AK033834.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P135003 NM_001008426.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 741145 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 224076 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P110505 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P135003 AK033834.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P135003 NM_001008426.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729100 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729109 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729113 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729114 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729117 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729119 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729131 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729097 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729105 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729106 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729102 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729125 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P110505 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135003 AK033834.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135003 NM_001008426.3 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P110505 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00901664 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00901653 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00657756 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00901667 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00901672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729179 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729113 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00120116 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729137 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729114 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729146 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729116 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729121 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729142 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729153 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729165 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729156 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729155 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729091 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729102 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729130 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729152 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729169 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729086 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729168 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00809545 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729106 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729158 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729138 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729100 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00695743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729122 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729160 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00428750 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00875178 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00563227 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729118 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729120 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729150 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729144 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729125 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729140 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00901657 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729119 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729128 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729090 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729127 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729159 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00129331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729141 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729131 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729088 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729124 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729162 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00428744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729107 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729109 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00901667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729161 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10350750 - -
          Affymetrix Mouse Gene 1.1 ST Array 10350749 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 585020 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101190619 - -
          Illumina mouse-6 v1.0 expression beadchip 101190619 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1234154 - -
          Illumina Sentrix mouse-6.v1 bead array 101190619 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101190619 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2962539 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2962539 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline