Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01298330_m1
          See other SLFN4 GE Assays ›
          Gene Symbol:
          Slfn4
          Gene Name:
          schlafen 4
          Gene Aliases:
          AI450778
          Chromosome Location:
          Chr.11: 83175171 - 83190216 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01298330_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slfn4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK137559.1 - 6 - 7 1351 63
          AK137689.1 - 5 - 6 1272 63
          BC044865.1 - 6 - 7 1360 4459564 63
          BC150770.1 - 3 - 4 967 9087681 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001241.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P183812 BC150770.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P183812 AK137689.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P183812 BC044865.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P183812 BC046511.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P183812 AF099976.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P183812 BC150770.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P183812 AK137689.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P183812 BC044865.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P183812 BC046511.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P183812 AF099976.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 692012 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 570009 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 93898 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 76520 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 832827 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 438459 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 874995 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 870420 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 106144 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 11645 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153808 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140963 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P183812 BC150770.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P183812 AK137689.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P183812 BC044865.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P183812 BC046511.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P183812 AF099976.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P183812 BC150770.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P183812 AK137689.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P183812 BC044865.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P183812 BC046511.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P183812 AF099976.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27970233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27970169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27970170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27970242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27970246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27970156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27970162 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27970231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27970232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27970166 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27970220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27970218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27970175 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27970161 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27970154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27970229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27970244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27970171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27970168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27970157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27970164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27970158 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27970167 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27970241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27970174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27970177 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27970153 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140963 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153808 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07420239 - -
          Whole Mouse Genome G4122A A_51_P183812 BC150770.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P183812 AK137689.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P183812 BC044865.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P183812 BC046511.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P183812 AF099976.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P183812 BC150770.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P183812 AK137689.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P183812 BC044865.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P183812 BC046511.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P183812 AF099976.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135213 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133836 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140963 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153808 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09339725 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09610742 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09339723 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09610748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27970269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09170158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09610748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09280812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09318773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09554508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09429361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09211843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09641411 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09007733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09554525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27970199 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10379642 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379637 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379644 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379643 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379645 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379639 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379641 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379640 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379638 - -
          Affymetrix HT MG-430 PM Array Plate 1427102_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10379636 - -
          Mouse Genome 430 2.0 Array 1427102_at - -
          Mouse Genome 430 2.0 Array 1427102_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20558_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20558_at - -
          Affymetrix Mouse Clariom S Array PSR1100011645.mm.1 NM_011410.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011645.mm.1 TC1100001241.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100011644.mm.1 NM_011410.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011644.mm.1 TC1100001241.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100011647.mm.1 NM_011410.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011647.mm.1 TC1100001241.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100011646.mm.1 NM_011410.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011646.mm.1 TC1100001241.mm.1 MTA TC
          Mouse Expression Set 430 1450322_PM_s_at - -
          Mouse Expression Set 430 1427102_PM_at - -
          Mouse Expression Set 430 1427102_at - -
          Mouse Expression Set 430 1450322_s_at - -
          Murine Genome U74v2 Set 92315_at - -
          Murine Genome U74v2 Set 98299_PM_s_at - -
          Murine Genome U74v2 Set 92315_PM_at - -
          Murine Genome U74v2 Set 98299_s_at - -
          Mouse Genome 430A 2.0 Array 1450322_s_at - -
          Mouse Genome 430A 2.0 Array 1450322_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1427102_at - -
          Mouse Genome 430A 2.0 Array 1427102_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427102_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427102_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 333745 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3190750 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003190750 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1222206 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222206 - -
          mouse-6 v1.1 (Illumina) scl41145.8.1_28-S - -
          Illumina Sentrix mouse-6.v1 bead array 3190750 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl41145.8.1_28-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3190750 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1222206 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1222206 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0