Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01302848_m1
          See other CCDC176 GE Assays ›
          Gene Symbol:
          Ccdc176
          Gene Name:
          coiled-coil domain containing 176
          Gene Aliases:
          2900006K08Rik, AI427784
          Chromosome Location:
          Chr.12: 84409068 - 84441763 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01302848_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccdc176
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028377.3 NP_082653.2 5 - 6 714 76
          GenBank mRNA AK133381.1 - 5 - 6 669 76
          BC151099.1 - 5 - 6 636 9088012 76
          BC151103.1 - 5 - 6 636 9088016 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200000810.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccdc176

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976494 NM_028377.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976494 AK133381.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 624075 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 135849 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 130576 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 119165 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 19192 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 182593 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 69074 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 505395 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 76313 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 631932 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 527405 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 219716 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 652511 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 575885 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 647352 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 81639 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 75514 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 93666 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119478 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163925 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976494 NM_028377.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976494 AK133381.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28598168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28598173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28598171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28598167 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28598172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28598166 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163925 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02565860 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P119478 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07690496 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07690560 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07690539 - -
          Whole Mouse Genome G4122A A_51_P171531 BC151099.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P171531 BC151103.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P171531 NM_028377.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P171531 AK013494.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P171531 AK133381.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P171533 BC151099.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P171533 BC151103.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P171533 NM_028377.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P171533 AK013494.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P171533 AK133381.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116374 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P119478 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07690539 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28598164 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28598180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28598196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28598195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28598191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28598173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28598188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28598198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28598185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28598179 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28598169 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28598168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28598171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28598190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28598167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28598193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28598172 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28598186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28598176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28598187 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28598183 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10397240 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397237 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397235 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397238 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397231 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397243 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397239 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397242 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397241 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397233 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397232 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397236 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397234 - -
          Affymetrix HT MG-430 PM Array Plate 1438122_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10397230 - -
          Mouse Genome 430 2.0 Array 1438122_at - -
          Mouse Genome 430 2.0 Array 1438122_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72873_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72873_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1200005862.mm.1 NM_028377.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200005862.mm.1 NM_028377.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200005862.mm.1 TC1200000810.mm.1 MTA TC
          Mouse Expression Set 430 1438122_PM_at - -
          Mouse Expression Set 430 1438122_at - -
          Affymetrix HT Mouse Genome 430B Array 1438122_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1438122_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 316701 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 107040110 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1229947 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2948800 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2948800 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline