Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01305934_m1
          See other HOXC11 GE Assays ›
          Gene Symbol:
          Hoxc11
          Gene Name:
          homeobox C11
          Gene Aliases:
          Hox-3.7
          Chromosome Location:
          Chr.15: 102954526 - 102956701 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01305934_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hoxc11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024842.1 NP_001020013.1 1 - 2 688 78
          GenBank mRNA BC139233.1 - 1 - 2 734 8862255 78
          BC139269.1 - 1 - 2 734 8862291 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500001116.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hoxc11

          Panther Classification:

          Molecular Function -

          homeodomain transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          metanephros development
          organ induction
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          multicellular organism development
          anterior/posterior pattern specification
          proximal/distal pattern formation
          embryonic digit morphogenesis
          positive regulation of transcription from RNA polymerase II promoter
          embryonic skeletal joint morphogenesis
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30500960 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30500978 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30500976 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30500959 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30500966 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30500968 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30500965 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30500962 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30500977 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30500964 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30500975 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2028399 BC139233.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2028399 BC139269.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2028399 NM_001024842.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 495639 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 684807 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2028399 BC139233.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2028399 BC139269.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2028399 NM_001024842.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500975 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500947 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500967 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500936 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500978 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500950 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500976 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500971 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500972 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500919 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500957 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500934 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500970 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30500925 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03119411 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30500964 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30500965 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30500959 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30500966 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30500977 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30500960 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30500968 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30500975 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30500976 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30500962 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30500978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30501015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30501024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30501033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30501036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30501047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30501016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30501037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30501018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30501040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30501020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30501044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30501041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30501034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30501058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30501032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30501030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30501042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30501050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30501054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30501029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30501021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30501027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30501045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30501025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30501055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30501052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30501038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30500962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11986373 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12077227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12296431 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11986382 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500973 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12012145 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500928 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12224401 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500932 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12212259 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12212268 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500906 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500895 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500929 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500920 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500930 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11986366 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12077225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11986386 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12296452 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500902 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12296441 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12296436 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12077223 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12012144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12296460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500956 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500945 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30500947 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10427279 - -
          Affymetrix Mouse Gene 1.0 ST Array 10427278 - -
          Affymetrix Mouse Gene 1.1 ST Array 10427277 - -
          Affymetrix Mouse Clariom S Array PSR1500008976.mm.1 NM_001024842.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500008976.mm.1 NM_001024842.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500008976.mm.1 TC1500001116.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500008974.mm.1 TC1500001116.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500008975.mm.1 NM_001024842.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500008975.mm.1 NM_001024842.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500008975.mm.1 TC1500001116.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500008977.mm.1 TC1500001116.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 350406 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1245642 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2988178 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2988178 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline